Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43824198-43824463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG5
Entrez Gene ID
64240
Gene Name
ATP binding cassette subfamily G member 5
Gene Aliases
STSL
UniGene
Hs.132992
Gene Chromosome Location
Chr.2: 43806154-43838840 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_006712074.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
UniGene
Hs.371597
Gene Chromosome Location
Chr.2: 43774039-43828492 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_006712074.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4658482 c.997G>A p.A333T Chr.2: 43824340-43824340 on Build GRCh38
384930 c.1038A>T p.K346N Chr.2: 43824299-43824299 on Build GRCh38
4583521 c.1049C>G p.T350R Chr.2: 43824288-43824288 on Build GRCh38
1614864 c.1117A>T p.R373W Chr.2: 43824220-43824220 on Build GRCh38
4166877 c.940C>T p.R314W Chr.2: 43824397-43824397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746632134 Chr.2: 43824224-43824224 on Build GRCh38
rs142037828 Chr.2: 43824434-43824435 on Build GRCh38
rs142971825 Chr.2: 43824397-43824397 on Build GRCh38
rs369775776 Chr.2: 43824368-43824368 on Build GRCh38
rs776365033 Chr.2: 43824255-43824255 on Build GRCh38
rs761733742 Chr.2: 43824408-43824408 on Build GRCh38
rs371319986 Chr.2: 43824293-43824293 on Build GRCh38
rs148131742 Chr.2: 43824288-43824288 on Build GRCh38
rs867011983 Chr.2: 43824274-43824274 on Build GRCh38
rs764153556 Chr.2: 43824283-43824283 on Build GRCh38
rs771202498 Chr.2: 43824345-43824347 on Build GRCh38
rs553464142 Chr.2: 43824338-43824338 on Build GRCh38
rs971595317 Chr.2: 43824228-43824228 on Build GRCh38
rs754395947 Chr.2: 43824341-43824341 on Build GRCh38
rs770535982 Chr.2: 43824227-43824227 on Build GRCh38
rs768533843 Chr.2: 43824401-43824401 on Build GRCh38
rs774304589 Chr.2: 43824404-43824404 on Build GRCh38
rs777402726 Chr.2: 43824363-43824363 on Build GRCh38
rs773296588 Chr.2: 43824422-43824422 on Build GRCh38
rs373926312 Chr.2: 43824417-43824417 on Build GRCh38
rs143740796 Chr.2: 43824423-43824423 on Build GRCh38
rs774717550 Chr.2: 43824419-43824462 on Build GRCh38
rs1014715108 Chr.2: 43824350-43824350 on Build GRCh38
rs867691050 Chr.2: 43824345-43824345 on Build GRCh38
rs374919194 Chr.2: 43824271-43824271 on Build GRCh38
rs767648807 Chr.2: 43824415-43824415 on Build GRCh38
rs766912458 Chr.2: 43824333-43824333 on Build GRCh38
rs868003691 Chr.2: 43824249-43824249 on Build GRCh38
rs866292096 Chr.2: 43824277-43824277 on Build GRCh38
rs762934862 Chr.2: 43824272-43824272 on Build GRCh38
rs756376137 Chr.2: 43824322-43824322 on Build GRCh38
rs368123646 Chr.2: 43824292-43824292 on Build GRCh38
rs751087839 Chr.2: 43824287-43824287 on Build GRCh38
rs750663975 Chr.2: 43824312-43824312 on Build GRCh38
rs146131711 Chr.2: 43824380-43824380 on Build GRCh38
rs775348848 Chr.2: 43824270-43824270 on Build GRCh38
rs869025350 Chr.2: 43824359-43824359 on Build GRCh38
rs544541406 Chr.2: 43824396-43824396 on Build GRCh38
rs745606275 Chr.2: 43824269-43824269 on Build GRCh38
rs754306032 Chr.2: 43824435-43824435 on Build GRCh38
rs749191249 Chr.2: 43824398-43824398 on Build GRCh38
rs201302611 Chr.2: 43824357-43824357 on Build GRCh38
rs780639375 Chr.2: 43824379-43824379 on Build GRCh38
rs374824368 Chr.2: 43824294-43824294 on Build GRCh38

View Full Product Details