Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136556360-136556633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VGLL1
Entrez Gene ID
51442
Gene Name
vestigial like family member 1
Gene Aliases
TDU, VGL1
UniGene
Hs.496843
Gene Chromosome Location
Chr.X: 136532152-136556807 on Build GRCh38
Transcripts
NM_016267.3

SNP Detail

SNP ID Chromosome Location
rs760707143 Chr.X: 136556482-136556482 on Build GRCh38
rs774814208 Chr.X: 136556483-136556484 on Build GRCh38
rs367554675 Chr.X: 136556591-136556591 on Build GRCh38
rs945106974 Chr.X: 136556539-136556539 on Build GRCh38
rs762602632 Chr.X: 136556485-136556488 on Build GRCh38
rs762245255 Chr.X: 136556452-136556452 on Build GRCh38
rs368763104 Chr.X: 136556570-136556570 on Build GRCh38
rs960309129 Chr.X: 136556434-136556434 on Build GRCh38
rs199963506 Chr.X: 136556401-136556401 on Build GRCh38
rs181846427 Chr.X: 136556384-136556384 on Build GRCh38
rs1042544296 Chr.X: 136556567-136556567 on Build GRCh38
rs762827354 Chr.X: 136556588-136556588 on Build GRCh38
rs749782510 Chr.X: 136556430-136556430 on Build GRCh38
rs3027868 Chr.X: 136556533-136556533 on Build GRCh38
rs993003903 Chr.X: 136556438-136556438 on Build GRCh38
rs772479081 Chr.X: 136556486-136556486 on Build GRCh38
rs375155236 Chr.X: 136556435-136556435 on Build GRCh38
rs913430811 Chr.X: 136556506-136556506 on Build GRCh38
rs1048831107 Chr.X: 136556412-136556412 on Build GRCh38
rs754064362 Chr.X: 136556563-136556563 on Build GRCh38
rs377351059 Chr.X: 136556405-136556405 on Build GRCh38
rs980328236 Chr.X: 136556381-136556381 on Build GRCh38
rs370933831 Chr.X: 136556574-136556574 on Build GRCh38
rs1038704446 Chr.X: 136556597-136556597 on Build GRCh38
rs763809306 Chr.X: 136556529-136556529 on Build GRCh38
rs112678338 Chr.X: 136556421-136556421 on Build GRCh38
rs770383296 Chr.X: 136556408-136556408 on Build GRCh38
rs769302209 Chr.X: 136556411-136556411 on Build GRCh38
rs373797445 Chr.X: 136556584-136556584 on Build GRCh38
rs769168051 Chr.X: 136556446-136556446 on Build GRCh38
rs774619886 Chr.X: 136556449-136556449 on Build GRCh38

View Full Product Details