Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58084621-58084866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN18
Entrez Gene ID
65982
Gene Name
zinc finger and SCAN domain containing 18
Gene Aliases
ZNF447
UniGene
Hs.235390
Gene Chromosome Location
Chr.19: 58083834-58118433 on Build GRCh38
Transcripts
NM_001145542.1 NM_001145543.1 NM_001145544.1 NM_023926.4 XM_005259174.4 XM_006723335.2 XM_011527237.2 XM_011527238.1 XM_011527239.2 XM_017027169.1 XM_017027170.1 XM_017027171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751218 c.1539C>T p.A513A Chr.19: 58084847-58084847 on Build GRCh38
6441062 c.1603C>T p.P535S Chr.19: 58084783-58084783 on Build GRCh38
6834256 c.1509C>T p.P503P Chr.19: 58084709-58084709 on Build GRCh38
5985083 c.1419C>G p.G473G Chr.19: 58084799-58084799 on Build GRCh38
4082314 c.1489G>A p.V497M Chr.19: 58084729-58084729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374324467 Chr.19: 58084829-58084829 on Build GRCh38
rs745750512 Chr.19: 58084642-58084642 on Build GRCh38
rs765241107 Chr.19: 58084808-58084808 on Build GRCh38
rs950899331 Chr.19: 58084668-58084668 on Build GRCh38
rs768334380 Chr.19: 58084690-58084690 on Build GRCh38
rs777378573 Chr.19: 58084748-58084748 on Build GRCh38
rs762185963 Chr.19: 58084784-58084784 on Build GRCh38
rs778827473 Chr.19: 58084696-58084696 on Build GRCh38
rs755160245 Chr.19: 58084710-58084736 on Build GRCh38
rs911144762 Chr.19: 58084781-58084781 on Build GRCh38
rs1031112677 Chr.19: 58084755-58084755 on Build GRCh38
rs775951653 Chr.19: 58084760-58084760 on Build GRCh38
rs777935727 Chr.19: 58084653-58084653 on Build GRCh38
rs974996794 Chr.19: 58084702-58084702 on Build GRCh38
rs886495865 Chr.19: 58084827-58084827 on Build GRCh38
rs1026091088 Chr.19: 58084811-58084811 on Build GRCh38
rs758710397 Chr.19: 58084648-58084648 on Build GRCh38
rs1013348446 Chr.19: 58084663-58084663 on Build GRCh38
rs749423179 Chr.19: 58084683-58084684 on Build GRCh38
rs764438118 Chr.19: 58084722-58084722 on Build GRCh38
rs200742278 Chr.19: 58084749-58084749 on Build GRCh38
rs779172201 Chr.19: 58084823-58084825 on Build GRCh38
rs751724875 Chr.19: 58084664-58084664 on Build GRCh38
rs529233652 Chr.19: 58084708-58084708 on Build GRCh38
rs1006916300 Chr.19: 58084681-58084681 on Build GRCh38
rs754830637 Chr.19: 58084800-58084800 on Build GRCh38
rs756944987 Chr.19: 58084838-58084838 on Build GRCh38
rs775144916 Chr.19: 58084739-58084739 on Build GRCh38
rs752680446 Chr.19: 58084816-58084816 on Build GRCh38
rs780530949 Chr.19: 58084848-58084848 on Build GRCh38
rs886174740 Chr.19: 58084822-58084822 on Build GRCh38
rs765672518 Chr.19: 58084646-58084646 on Build GRCh38
rs759793951 Chr.19: 58084643-58084643 on Build GRCh38
rs200522523 Chr.19: 58084797-58084797 on Build GRCh38
rs774710516 Chr.19: 58084773-58084773 on Build GRCh38
rs200894425 Chr.19: 58084798-58084798 on Build GRCh38
rs201438629 Chr.19: 58084710-58084710 on Build GRCh38
rs146022713 Chr.19: 58084742-58084742 on Build GRCh38
rs367790148 Chr.19: 58084745-58084745 on Build GRCh38
rs779834219 Chr.19: 58084673-58084673 on Build GRCh38
rs780214894 Chr.19: 58084669-58084669 on Build GRCh38
rs757996966 Chr.19: 58084746-58084746 on Build GRCh38
rs772176251 Chr.19: 58084704-58084704 on Build GRCh38
rs199767535 Chr.19: 58084767-58084767 on Build GRCh38
rs757381871 Chr.19: 58084724-58084724 on Build GRCh38
rs747738797 Chr.19: 58084697-58084697 on Build GRCh38
rs751879810 Chr.19: 58084723-58084723 on Build GRCh38
rs748525601 Chr.19: 58084675-58084675 on Build GRCh38
rs886424628 Chr.19: 58084669-58084669 on Build GRCh38
rs922438695 Chr.19: 58084806-58084806 on Build GRCh38
rs985343636 Chr.19: 58084671-58084671 on Build GRCh38
rs771715014 Chr.19: 58084699-58084699 on Build GRCh38
rs534784715 Chr.19: 58084744-58084744 on Build GRCh38
rs770519899 Chr.19: 58084707-58084707 on Build GRCh38
rs753113170 Chr.19: 58084714-58084714 on Build GRCh38
rs569550361 Chr.19: 58084701-58084701 on Build GRCh38
rs763230938 Chr.19: 58084721-58084721 on Build GRCh38
rs1033777424 Chr.19: 58084645-58084645 on Build GRCh38
rs143846640 Chr.19: 58084826-58084826 on Build GRCh38
rs752855294 Chr.19: 58084647-58084647 on Build GRCh38
rs746647847 Chr.19: 58084832-58084832 on Build GRCh38
rs1016532250 Chr.19: 58084682-58084682 on Build GRCh38
rs781401678 Chr.19: 58084725-58084725 on Build GRCh38
rs763391279 Chr.19: 58084762-58084762 on Build GRCh38
rs760153250 Chr.19: 58084709-58084709 on Build GRCh38
rs758154343 Chr.19: 58084817-58084817 on Build GRCh38
rs549791527 Chr.19: 58084684-58084684 on Build GRCh38
rs756082128 Chr.19: 58084666-58084667 on Build GRCh38
rs745409018 Chr.19: 58084849-58084849 on Build GRCh38
rs761487636 Chr.19: 58084757-58084757 on Build GRCh38
rs570885021 Chr.19: 58084756-58084756 on Build GRCh38
rs756069756 Chr.19: 58084666-58084666 on Build GRCh38

View Full Product Details