Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32378086-32378317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSDC1
Entrez Gene ID
55108
Gene Name
BSD domain containing 1
Gene Aliases
-
UniGene
Hs.353454
Gene Chromosome Location
Chr.1: 32364633-32394461 on Build GRCh38
Transcripts
NM_001143888.2 NM_001143889.2 NM_001143890.2 NM_001300958.1 NM_018045.7 NR_125352.1 XM_005270986.4 XM_011541677.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3488175 c.609C>T p.F203F Chr.1: 32378254-32378254 on Build GRCh38
6291966 c.580-7C>T p.? Chr.1: 32378290-32378290 on Build GRCh38
6291967 c.580-7C>T p.? Chr.1: 32378290-32378290 on Build GRCh38
7411910 c.617G>A p.R206Q Chr.1: 32378246-32378246 on Build GRCh38
3488176 c.609C>T p.F203F Chr.1: 32378254-32378254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766366862 Chr.1: 32378228-32378228 on Build GRCh38
rs760817406 Chr.1: 32378164-32378164 on Build GRCh38
rs765632276 Chr.1: 32378181-32378181 on Build GRCh38
rs750992561 Chr.1: 32378182-32378182 on Build GRCh38
rs377336946 Chr.1: 32378294-32378294 on Build GRCh38
rs373057622 Chr.1: 32378233-32378233 on Build GRCh38
rs150963407 Chr.1: 32378227-32378227 on Build GRCh38
rs979728441 Chr.1: 32378237-32378237 on Build GRCh38
rs560058496 Chr.1: 32378289-32378289 on Build GRCh38
rs577793168 Chr.1: 32378184-32378184 on Build GRCh38
rs867979679 Chr.1: 32378254-32378254 on Build GRCh38
rs777450364 Chr.1: 32378247-32378247 on Build GRCh38
rs764013404 Chr.1: 32378292-32378293 on Build GRCh38
rs751121688 Chr.1: 32378220-32378220 on Build GRCh38
rs144659572 Chr.1: 32378280-32378280 on Build GRCh38
rs760803134 Chr.1: 32378287-32378287 on Build GRCh38
rs770858246 Chr.1: 32378267-32378267 on Build GRCh38
rs749183364 Chr.1: 32378260-32378260 on Build GRCh38
rs762062939 Chr.1: 32378291-32378291 on Build GRCh38
rs779673645 Chr.1: 32378185-32378185 on Build GRCh38
rs754661592 Chr.1: 32378223-32378223 on Build GRCh38
rs866514828 Chr.1: 32378271-32378271 on Build GRCh38
rs562590616 Chr.1: 32378135-32378135 on Build GRCh38
rs374773298 Chr.1: 32378177-32378177 on Build GRCh38
rs765592300 Chr.1: 32378293-32378293 on Build GRCh38
rs768863124 Chr.1: 32378288-32378288 on Build GRCh38
rs992779914 Chr.1: 32378196-32378196 on Build GRCh38
rs757708121 Chr.1: 32378179-32378179 on Build GRCh38

View Full Product Details