Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 21623677-21623916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRPD1
Entrez Gene ID
6632
Gene Name
small nuclear ribonucleoprotein D1 polypeptide
Gene Aliases
HsT2456, SMD1, SNRPD, Sm-D1
UniGene
Hs.464734
Gene Chromosome Location
Chr.18: 21612269-21630247 on Build GRCh38
Transcripts
NM_001291916.1 NM_006938.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7124463 c.182G>C p.R61P Chr.18: 21623838-21623838 on Build GRCh38
393339 c.198G>C p.R66R Chr.18: 21623854-21623854 on Build GRCh38
986945 c.182G>A p.R61Q Chr.18: 21623838-21623838 on Build GRCh38
4071445 c.171G>A p.T57T Chr.18: 21623827-21623827 on Build GRCh38
6862448 c.92G>C p.G31A Chr.18: 21623748-21623748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746642432 Chr.18: 21623814-21623814 on Build GRCh38
rs376513859 Chr.18: 21623720-21623720 on Build GRCh38
rs559622247 Chr.18: 21623839-21623839 on Build GRCh38
rs751716137 Chr.18: 21623717-21623717 on Build GRCh38
rs777542633 Chr.18: 21623811-21623811 on Build GRCh38
rs757844839 Chr.18: 21623745-21623745 on Build GRCh38
rs958892922 Chr.18: 21623810-21623810 on Build GRCh38
rs754927231 Chr.18: 21623731-21623731 on Build GRCh38
rs11548769 Chr.18: 21623777-21623777 on Build GRCh38
rs775855047 Chr.18: 21623890-21623890 on Build GRCh38
rs778655238 Chr.18: 21623742-21623742 on Build GRCh38
rs771463023 Chr.18: 21623827-21623827 on Build GRCh38
rs764442658 Chr.18: 21623709-21623709 on Build GRCh38
rs766130152 Chr.18: 21623723-21623723 on Build GRCh38
rs760760064 Chr.18: 21623722-21623722 on Build GRCh38
rs141758776 Chr.18: 21623866-21623866 on Build GRCh38
rs572142783 Chr.18: 21623852-21623852 on Build GRCh38
rs201620624 Chr.18: 21623724-21623724 on Build GRCh38

View Full Product Details