Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42194715-42194984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAK1
Entrez Gene ID
22906
Gene Name
trafficking kinesin protein 1
Gene Aliases
MILT1, OIP106
UniGene
Hs.535711
Gene Chromosome Location
Chr.3: 42013802-42225890 on Build GRCh38
Transcripts
NM_001042646.2 NM_001265608.1 NM_001265609.1 NM_001265610.1 NM_014965.4 XM_005264956.2 XM_005264958.2 XM_005264960.3 XM_005264962.3 XM_005264963.3 XM_006713029.2 XM_006713030.2 XM_006713031.3 XM_011533489.2 XM_017005906.1 XM_017005907.1 XM_017005908.1 XM_017005909.1 XM_017005910.1 XM_017005911.1 XM_017005912.1 XM_017005913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459028 c.1109C>T p.P370L Chr.3: 42194937-42194937 on Build GRCh38
4459011 c.1108C>T p.P370S Chr.3: 42194936-42194936 on Build GRCh38
4459010 c.934C>T p.P312S Chr.3: 42194936-42194936 on Build GRCh38
2988169 c.1084C>T p.R362C Chr.3: 42194912-42194912 on Build GRCh38
4459027 c.935C>T p.P312L Chr.3: 42194937-42194937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184870601 Chr.3: 42194965-42194965 on Build GRCh38
rs746535309 Chr.3: 42194909-42194909 on Build GRCh38
rs987339098 Chr.3: 42194961-42194961 on Build GRCh38
rs761614570 Chr.3: 42194857-42194857 on Build GRCh38
rs758939739 Chr.3: 42194830-42194830 on Build GRCh38
rs748462432 Chr.3: 42194808-42194808 on Build GRCh38
rs374334482 Chr.3: 42194922-42194922 on Build GRCh38
rs773021871 Chr.3: 42194920-42194920 on Build GRCh38
rs370769044 Chr.3: 42194769-42194769 on Build GRCh38
rs760439072 Chr.3: 42194760-42194760 on Build GRCh38
rs774989973 Chr.3: 42194838-42194838 on Build GRCh38
rs761561436 Chr.3: 42194913-42194913 on Build GRCh38
rs549322574 Chr.3: 42194856-42194856 on Build GRCh38
rs1032437486 Chr.3: 42194735-42194735 on Build GRCh38
rs756293488 Chr.3: 42194929-42194929 on Build GRCh38
rs770281448 Chr.3: 42194814-42194814 on Build GRCh38
rs755001607 Chr.3: 42194860-42194860 on Build GRCh38
rs751540230 Chr.3: 42194789-42194789 on Build GRCh38
rs144710739 Chr.3: 42194939-42194939 on Build GRCh38
rs954387168 Chr.3: 42194775-42194775 on Build GRCh38
rs749688177 Chr.3: 42194821-42194821 on Build GRCh38
rs72869992 Chr.3: 42194751-42194751 on Build GRCh38
rs757737865 Chr.3: 42194902-42194902 on Build GRCh38
rs754163195 Chr.3: 42194964-42194964 on Build GRCh38
rs575394877 Chr.3: 42194820-42194820 on Build GRCh38
rs569323766 Chr.3: 42194867-42194867 on Build GRCh38
rs763740116 Chr.3: 42194851-42194851 on Build GRCh38
rs771664475 Chr.3: 42194837-42194837 on Build GRCh38
rs749630929 Chr.3: 42194898-42194898 on Build GRCh38
rs752728388 Chr.3: 42194878-42194878 on Build GRCh38
rs771516117 Chr.3: 42194828-42194828 on Build GRCh38
rs768244951 Chr.3: 42194910-42194910 on Build GRCh38
rs1017687007 Chr.3: 42194834-42194834 on Build GRCh38
rs752436789 Chr.3: 42194754-42194754 on Build GRCh38
rs914460522 Chr.3: 42194919-42194919 on Build GRCh38
rs763934196 Chr.3: 42194761-42194761 on Build GRCh38
rs779267051 Chr.3: 42194904-42194904 on Build GRCh38
rs771476359 Chr.3: 42194907-42194907 on Build GRCh38
rs370668547 Chr.3: 42194879-42194879 on Build GRCh38
rs532911279 Chr.3: 42194850-42194850 on Build GRCh38
rs753770039 Chr.3: 42194766-42194766 on Build GRCh38
rs778163685 Chr.3: 42194884-42194884 on Build GRCh38
rs746442830 Chr.3: 42194905-42194905 on Build GRCh38
rs139261502 Chr.3: 42194901-42194901 on Build GRCh38
rs373149810 Chr.3: 42194796-42194796 on Build GRCh38
rs144731827 Chr.3: 42194859-42194859 on Build GRCh38
rs756350710 Chr.3: 42194880-42194880 on Build GRCh38
rs376331284 Chr.3: 42194798-42194798 on Build GRCh38
rs765126555 Chr.3: 42194917-42194917 on Build GRCh38
rs951812410 Chr.3: 42194827-42194827 on Build GRCh38
rs778830600 Chr.3: 42194780-42194780 on Build GRCh38
rs776292575 Chr.3: 42194912-42194912 on Build GRCh38
rs9851455 Chr.3: 42194923-42194923 on Build GRCh38
rs757570416 Chr.3: 42194734-42194734 on Build GRCh38

View Full Product Details