Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148741376-148741649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTR1
Entrez Gene ID
185
Gene Name
angiotensin II receptor type 1
Gene Aliases
AG2S, AGTR1B, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, HAT1R
UniGene
Hs.477887
Gene Chromosome Location
Chr.3: 148697871-148743003 on Build GRCh38
Transcripts
NM_000685.4 NM_004835.4 NM_009585.3 NM_031850.3 NM_032049.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
387374 c.359C>A p.T120K Chr.3: 148741394-148741394 on Build GRCh38
1419804 c.513C>G p.F171L Chr.3: 148741548-148741548 on Build GRCh38
728841 c.385G>T p.A129S Chr.3: 148741420-148741420 on Build GRCh38
3944980 c.422C>A p.T141K Chr.3: 148741457-148741457 on Build GRCh38
3846462 c.377G>A p.R126Q Chr.3: 148741412-148741412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995803867 Chr.3: 148741444-148741444 on Build GRCh38
rs369560166 Chr.3: 148741570-148741570 on Build GRCh38
rs917905020 Chr.3: 148741600-148741600 on Build GRCh38
rs763952495 Chr.3: 148741453-148741453 on Build GRCh38
rs757886940 Chr.3: 148741406-148741406 on Build GRCh38
rs369846514 Chr.3: 148741435-148741435 on Build GRCh38
rs766887107 Chr.3: 148741543-148741544 on Build GRCh38
rs780264737 Chr.3: 148741501-148741501 on Build GRCh38
rs56257794 Chr.3: 148741445-148741445 on Build GRCh38
rs778815388 Chr.3: 148741464-148741464 on Build GRCh38
rs202211101 Chr.3: 148741562-148741562 on Build GRCh38
rs979012840 Chr.3: 148741552-148741552 on Build GRCh38
rs773829709 Chr.3: 148741429-148741429 on Build GRCh38
rs1038279926 Chr.3: 148741616-148741616 on Build GRCh38
rs968555821 Chr.3: 148741505-148741505 on Build GRCh38
rs763613654 Chr.3: 148741591-148741591 on Build GRCh38
rs761847680 Chr.3: 148741601-148741601 on Build GRCh38
rs553350297 Chr.3: 148741588-148741588 on Build GRCh38
rs772616879 Chr.3: 148741610-148741610 on Build GRCh38
rs570056677 Chr.3: 148741454-148741454 on Build GRCh38
rs760594254 Chr.3: 148741451-148741451 on Build GRCh38
rs770227887 Chr.3: 148741415-148741415 on Build GRCh38
rs775127751 Chr.3: 148741585-148741585 on Build GRCh38
rs766907479 Chr.3: 148741442-148741442 on Build GRCh38
rs1020069665 Chr.3: 148741481-148741481 on Build GRCh38
rs200184769 Chr.3: 148741535-148741535 on Build GRCh38
rs762020328 Chr.3: 148741412-148741412 on Build GRCh38
rs768866306 Chr.3: 148741534-148741534 on Build GRCh38
rs949413715 Chr.3: 148741604-148741604 on Build GRCh38
rs868647200 Chr.3: 148741581-148741581 on Build GRCh38
rs764933525 Chr.3: 148741606-148741606 on Build GRCh38
rs748715507 Chr.3: 148741550-148741550 on Build GRCh38
rs5182 Chr.3: 148741608-148741608 on Build GRCh38
rs373754918 Chr.3: 148741617-148741617 on Build GRCh38
rs533866995 Chr.3: 148741564-148741564 on Build GRCh38
rs12721226 Chr.3: 148741522-148741522 on Build GRCh38
rs750577589 Chr.3: 148741470-148741470 on Build GRCh38
rs758709065 Chr.3: 148741479-148741479 on Build GRCh38
rs756951904 Chr.3: 148741462-148741462 on Build GRCh38
rs770034589 Chr.3: 148741560-148741560 on Build GRCh38
rs368534001 Chr.3: 148741405-148741405 on Build GRCh38
rs200844345 Chr.3: 148741597-148741597 on Build GRCh38
rs397514687 Chr.3: 148741411-148741411 on Build GRCh38
rs766645736 Chr.3: 148741611-148741611 on Build GRCh38
rs942053395 Chr.3: 148741512-148741512 on Build GRCh38
rs983478539 Chr.3: 148741605-148741605 on Build GRCh38
rs771771783 Chr.3: 148741584-148741584 on Build GRCh38
rs1038081292 Chr.3: 148741542-148741542 on Build GRCh38

View Full Product Details