Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58063399-58063672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF135
Entrez Gene ID
7694
Gene Name
zinc finger protein 135
Gene Aliases
ZNF61, ZNF78L1, pHZ-17, pT3
UniGene
Hs.85863
Gene Chromosome Location
Chr.19: 58059227-58069755 on Build GRCh38
Transcripts
NM_001164527.1 NM_001164529.1 NM_001164530.1 NM_001289401.1 NM_001289402.1 NM_003436.3 NM_007134.1 XM_005259211.1 XM_006723362.3 XM_006723363.3 XM_017027240.1 XM_017027241.1 XM_017027242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6558462 c.255G>T p.W85C Chr.19: 58063504-58063504 on Build GRCh38
4082294 c.246C>T p.G82G Chr.19: 58063531-58063531 on Build GRCh38
6558463 c.219G>T p.W73C Chr.19: 58063504-58063504 on Build GRCh38
1713027 c.191C>T p.S64F Chr.19: 58063476-58063476 on Build GRCh38
1713026 c.227C>T p.S76F Chr.19: 58063476-58063476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372228366 Chr.19: 58063541-58063541 on Build GRCh38
rs779229543 Chr.19: 58063478-58063478 on Build GRCh38
rs375491279 Chr.19: 58063459-58063459 on Build GRCh38
rs780298550 Chr.19: 58063605-58063605 on Build GRCh38
rs377476939 Chr.19: 58063582-58063582 on Build GRCh38
rs778062396 Chr.19: 58063560-58063560 on Build GRCh38
rs199686405 Chr.19: 58063464-58063464 on Build GRCh38
rs752061656 Chr.19: 58063614-58063617 on Build GRCh38
rs778310684 Chr.19: 58063596-58063596 on Build GRCh38
rs373371088 Chr.19: 58063574-58063574 on Build GRCh38
rs760843413 Chr.19: 58063430-58063430 on Build GRCh38
rs779431582 Chr.19: 58063542-58063542 on Build GRCh38
rs755663365 Chr.19: 58063465-58063465 on Build GRCh38
rs771035425 Chr.19: 58063501-58063501 on Build GRCh38
rs191053293 Chr.19: 58063461-58063461 on Build GRCh38
rs752403961 Chr.19: 58063523-58063523 on Build GRCh38
rs778065252 Chr.19: 58063493-58063493 on Build GRCh38
rs577329504 Chr.19: 58063481-58063481 on Build GRCh38
rs532596603 Chr.19: 58063611-58063611 on Build GRCh38
rs200386480 Chr.19: 58063467-58063467 on Build GRCh38
rs754339491 Chr.19: 58063476-58063476 on Build GRCh38
rs761703991 Chr.19: 58063510-58063510 on Build GRCh38
rs781338674 Chr.19: 58063537-58063537 on Build GRCh38
rs780494007 Chr.19: 58063442-58063442 on Build GRCh38
rs774248199 Chr.19: 58063508-58063508 on Build GRCh38
rs747232800 Chr.19: 58063615-58063615 on Build GRCh38
rs747249059 Chr.19: 58063496-58063496 on Build GRCh38
rs371178818 Chr.19: 58063458-58063458 on Build GRCh38
rs113591395 Chr.19: 58063456-58063456 on Build GRCh38
rs745975540 Chr.19: 58063603-58063603 on Build GRCh38
rs369565894 Chr.19: 58063559-58063559 on Build GRCh38
rs760313854 Chr.19: 58063532-58063532 on Build GRCh38
rs531203528 Chr.19: 58063432-58063432 on Build GRCh38
rs963108073 Chr.19: 58063632-58063632 on Build GRCh38
rs781370775 Chr.19: 58063584-58063584 on Build GRCh38
rs202077210 Chr.19: 58063437-58063437 on Build GRCh38
rs367734449 Chr.19: 58063438-58063438 on Build GRCh38
rs376604339 Chr.19: 58063557-58063557 on Build GRCh38
rs758619510 Chr.19: 58063556-58063557 on Build GRCh38
rs755521855 Chr.19: 58063477-58063477 on Build GRCh38
rs762960744 Chr.19: 58063507-58063507 on Build GRCh38
rs763947522 Chr.19: 58063447-58063447 on Build GRCh38
rs755022776 Chr.19: 58063435-58063436 on Build GRCh38
rs774444507 Chr.19: 58063643-58063643 on Build GRCh38
rs776610394 Chr.19: 58063506-58063506 on Build GRCh38
rs748538471 Chr.19: 58063480-58063480 on Build GRCh38
rs752997240 Chr.19: 58063554-58063554 on Build GRCh38
rs200161186 Chr.19: 58063523-58063523 on Build GRCh38
rs750077100 Chr.19: 58063531-58063531 on Build GRCh38
rs372275276 Chr.19: 58063419-58063419 on Build GRCh38
rs996328680 Chr.19: 58063492-58063492 on Build GRCh38

View Full Product Details