Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11965264-11965525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF17
Entrez Gene ID
608
Gene Name
TNF receptor superfamily member 17
Gene Aliases
BCM, BCMA, CD269, TNFRSF13A
UniGene
Hs.2556
Gene Chromosome Location
Chr.16: 11965107-11968068 on Build GRCh38
Transcripts
NM_001192.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749406 c.26C>T p.S9F Chr.16: 11965350-11965350 on Build GRCh38
3505854 c.37T>A p.Y13N Chr.16: 11965361-11965361 on Build GRCh38
4734875 c.80G>A p.R27Q Chr.16: 11965404-11965404 on Build GRCh38
7139403 c.73C>A p.Q25K Chr.16: 11965397-11965397 on Build GRCh38
7110015 c.34G>C p.E12Q Chr.16: 11965358-11965358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771579251 Chr.16: 11965419-11965421 on Build GRCh38
rs771330300 Chr.16: 11965453-11965456 on Build GRCh38
rs754875749 Chr.16: 11965347-11965347 on Build GRCh38
rs774030022 Chr.16: 11965438-11965438 on Build GRCh38
rs772767687 Chr.16: 11965400-11965400 on Build GRCh38
rs909750928 Chr.16: 11965474-11965474 on Build GRCh38
rs1013020007 Chr.16: 11965358-11965358 on Build GRCh38
rs767384244 Chr.16: 11965349-11965349 on Build GRCh38
rs372620061 Chr.16: 11965320-11965320 on Build GRCh38
rs200779776 Chr.16: 11965404-11965404 on Build GRCh38
rs760826489 Chr.16: 11965307-11965307 on Build GRCh38
rs756893990 Chr.16: 11965386-11965386 on Build GRCh38
rs143690193 Chr.16: 11965382-11965382 on Build GRCh38
rs118120349 Chr.16: 11965421-11965421 on Build GRCh38
rs752610824 Chr.16: 11965440-11965440 on Build GRCh38
rs144402082 Chr.16: 11965471-11965471 on Build GRCh38
rs751325020 Chr.16: 11965346-11965346 on Build GRCh38
rs763625636 Chr.16: 11965458-11965458 on Build GRCh38
rs369340296 Chr.16: 11965381-11965381 on Build GRCh38
rs778618861 Chr.16: 11965468-11965468 on Build GRCh38
rs201329134 Chr.16: 11965465-11965465 on Build GRCh38
rs550606391 Chr.16: 11965439-11965439 on Build GRCh38
rs753323763 Chr.16: 11965459-11965459 on Build GRCh38
rs746189405 Chr.16: 11965393-11965393 on Build GRCh38
rs749634242 Chr.16: 11965426-11965428 on Build GRCh38
rs1003746579 Chr.16: 11965389-11965389 on Build GRCh38
rs375886084 Chr.16: 11965379-11965379 on Build GRCh38
rs185085849 Chr.16: 11965403-11965403 on Build GRCh38
rs559017949 Chr.16: 11965388-11965388 on Build GRCh38
rs1036743739 Chr.16: 11965486-11965486 on Build GRCh38
rs369612176 Chr.16: 11965470-11965470 on Build GRCh38
rs752221019 Chr.16: 11965384-11965384 on Build GRCh38
rs11570140 Chr.16: 11965475-11965475 on Build GRCh38
rs190015454 Chr.16: 11965412-11965412 on Build GRCh38
rs760559467 Chr.16: 11965451-11965451 on Build GRCh38
rs762853349 Chr.16: 11965333-11965333 on Build GRCh38
rs976760993 Chr.16: 11965434-11965434 on Build GRCh38
rs766104779 Chr.16: 11965335-11965335 on Build GRCh38
rs747538009 Chr.16: 11965488-11965488 on Build GRCh38
rs775728959 Chr.16: 11965303-11965303 on Build GRCh38
rs138231933 Chr.16: 11965357-11965357 on Build GRCh38

View Full Product Details