Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33260990-33261263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG1
Entrez Gene ID
573
Gene Name
BCL2 associated athanogene 1
Gene Aliases
BAG-1, HAP, RAP46
UniGene
Hs.377484
Gene Chromosome Location
Chr.9: 33252471-33264761 on Build GRCh38
Transcripts
NM_001172415.1 NM_004323.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3782228 c.298G>A p.V100I Chr.9: 33261107-33261107 on Build GRCh38
5876118 c.296G>A p.R99Q Chr.9: 33261109-33261109 on Build GRCh38
5876117 c.641G>A p.R214Q Chr.9: 33261109-33261109 on Build GRCh38
6335402 c.581-9T>C p.? Chr.9: 33261178-33261178 on Build GRCh38
4665005 c.313A>G p.K105E Chr.9: 33261092-33261092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758738877 Chr.9: 33261122-33261122 on Build GRCh38
rs770917998 Chr.9: 33261132-33261132 on Build GRCh38
rs777155453 Chr.9: 33261192-33261192 on Build GRCh38
rs200190801 Chr.9: 33261083-33261083 on Build GRCh38
rs752936421 Chr.9: 33261120-33261120 on Build GRCh38
rs145328400 Chr.9: 33261094-33261094 on Build GRCh38
rs749670029 Chr.9: 33261107-33261107 on Build GRCh38
rs761326880 Chr.9: 33261180-33261180 on Build GRCh38
rs778036655 Chr.9: 33261058-33261058 on Build GRCh38
rs367846351 Chr.9: 33261159-33261159 on Build GRCh38
rs761083834 Chr.9: 33261100-33261100 on Build GRCh38
rs201889975 Chr.9: 33261124-33261124 on Build GRCh38
rs775088702 Chr.9: 33261085-33261085 on Build GRCh38
rs772237593 Chr.9: 33261054-33261054 on Build GRCh38
rs746879328 Chr.9: 33261186-33261186 on Build GRCh38
rs916746496 Chr.9: 33261031-33261031 on Build GRCh38
rs754296866 Chr.9: 33261046-33261046 on Build GRCh38
rs779216485 Chr.9: 33261048-33261048 on Build GRCh38
rs762637931 Chr.9: 33261091-33261091 on Build GRCh38
rs1008247404 Chr.9: 33261208-33261208 on Build GRCh38
rs371553280 Chr.9: 33261072-33261072 on Build GRCh38
rs981411197 Chr.9: 33261210-33261210 on Build GRCh38
rs914514390 Chr.9: 33261010-33261010 on Build GRCh38
rs746893690 Chr.9: 33261063-33261063 on Build GRCh38
rs199556100 Chr.9: 33261113-33261113 on Build GRCh38
rs948617468 Chr.9: 33261025-33261025 on Build GRCh38
rs760160265 Chr.9: 33261209-33261209 on Build GRCh38
rs766845235 Chr.9: 33261104-33261104 on Build GRCh38
rs150615545 Chr.9: 33261141-33261141 on Build GRCh38
rs745826979 Chr.9: 33261142-33261142 on Build GRCh38
rs766956598 Chr.9: 33261187-33261187 on Build GRCh38
rs953722695 Chr.9: 33261176-33261176 on Build GRCh38
rs779450407 Chr.9: 33261117-33261117 on Build GRCh38
rs200482625 Chr.9: 33261078-33261078 on Build GRCh38
rs762582550 Chr.9: 33261152-33261152 on Build GRCh38
rs770957327 Chr.9: 33261065-33261065 on Build GRCh38
rs768365765 Chr.9: 33261155-33261155 on Build GRCh38
rs748402691 Chr.9: 33261049-33261049 on Build GRCh38
rs376508916 Chr.9: 33261109-33261109 on Build GRCh38
rs755319430 Chr.9: 33261110-33261110 on Build GRCh38
rs751128311 Chr.9: 33261096-33261096 on Build GRCh38
rs373698859 Chr.9: 33261126-33261126 on Build GRCh38
rs578065772 Chr.9: 33261047-33261047 on Build GRCh38
rs775314108 Chr.9: 33261150-33261150 on Build GRCh38
rs753669035 Chr.9: 33261182-33261182 on Build GRCh38

View Full Product Details