Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 1052024-1052265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRT2
Entrez Gene ID
10655
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
UniGene
Hs.59506
Gene Chromosome Location
Chr.9: 1003237-1057554 on Build GRCh38
Transcripts
NM_001130865.2 NM_006557.6 NM_181872.4 XM_011517687.1 XM_011517690.2 XM_011517694.2 XM_017014213.1 XM_017014214.1 XM_017014215.1 XM_017014216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
396318 c.505C>A p.R169R Chr.9: 1052118-1052118 on Build GRCh38
7364646 c.436G>A p.D146N Chr.9: 1052049-1052049 on Build GRCh38
7364647 c.436G>A p.D146N Chr.9: 1052049-1052049 on Build GRCh38
396317 c.505C>A p.R169R Chr.9: 1052118-1052118 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758127591 Chr.9: 1052210-1052210 on Build GRCh38
rs1047495629 Chr.9: 1052214-1052214 on Build GRCh38
rs1031112044 Chr.9: 1052159-1052159 on Build GRCh38
rs958005260 Chr.9: 1052197-1052197 on Build GRCh38
rs906592565 Chr.9: 1052109-1052109 on Build GRCh38
rs1037914010 Chr.9: 1052048-1052048 on Build GRCh38
rs989516692 Chr.9: 1052201-1052201 on Build GRCh38
rs974581308 Chr.9: 1052221-1052221 on Build GRCh38
rs886208578 Chr.9: 1052218-1052218 on Build GRCh38
rs1012335797 Chr.9: 1052085-1052085 on Build GRCh38
rs566963920 Chr.9: 1052120-1052120 on Build GRCh38
rs535853385 Chr.9: 1052195-1052195 on Build GRCh38
rs928176686 Chr.9: 1052135-1052136 on Build GRCh38
rs1025567006 Chr.9: 1052087-1052087 on Build GRCh38
rs958069794 Chr.9: 1052072-1052072 on Build GRCh38
rs945516039 Chr.9: 1052131-1052131 on Build GRCh38
rs369093745 Chr.9: 1052061-1052061 on Build GRCh38
rs555991509 Chr.9: 1052205-1052205 on Build GRCh38
rs758230891 Chr.9: 1052056-1052056 on Build GRCh38
rs756386682 Chr.9: 1052108-1052108 on Build GRCh38
rs1009119554 Chr.9: 1052219-1052219 on Build GRCh38
rs982319193 Chr.9: 1052135-1052135 on Build GRCh38
rs780258369 Chr.9: 1052171-1052171 on Build GRCh38
rs1055424683 Chr.9: 1052158-1052158 on Build GRCh38
rs1000029198 Chr.9: 1052153-1052153 on Build GRCh38
rs938281882 Chr.9: 1052149-1052149 on Build GRCh38
rs1021519368 Chr.9: 1052208-1052208 on Build GRCh38
rs546986410 Chr.9: 1052092-1052092 on Build GRCh38
rs898826652 Chr.9: 1052189-1052189 on Build GRCh38
rs372312394 Chr.9: 1052075-1052075 on Build GRCh38
rs923110986 Chr.9: 1052225-1052225 on Build GRCh38
rs1018788028 Chr.9: 1052226-1052226 on Build GRCh38

View Full Product Details