Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31672131-31672392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN1
Entrez Gene ID
196394
Gene Name
antagonist of mitotic exit network 1 homolog
Gene Aliases
-
UniGene
Hs.585084
Gene Chromosome Location
Chr.12: 31671137-31729178 on Build GRCh38
Transcripts
NM_001113402.1 NM_001278411.1 NM_001278412.1 NR_103522.1 NR_103523.1 XM_006719054.3 XM_011520604.2 XM_017018963.1 XM_017018964.1 XM_017018965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1638212 c.746A>G p.K249R Chr.12: 31672335-31672335 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs578248025 Chr.12: 31672337-31672337 on Build GRCh38
rs11051524 Chr.12: 31672351-31672351 on Build GRCh38
rs774941640 Chr.12: 31672297-31672297 on Build GRCh38
rs765102110 Chr.12: 31672285-31672286 on Build GRCh38
rs115100488 Chr.12: 31672360-31672360 on Build GRCh38
rs950724925 Chr.12: 31672252-31672252 on Build GRCh38
rs1028052510 Chr.12: 31672242-31672242 on Build GRCh38
rs535205885 Chr.12: 31672240-31672241 on Build GRCh38
rs1006039318 Chr.12: 31672356-31672356 on Build GRCh38
rs773248890 Chr.12: 31672295-31672295 on Build GRCh38
rs1033766461 Chr.12: 31672184-31672188 on Build GRCh38
rs11551368 Chr.12: 31672186-31672186 on Build GRCh38
rs762340802 Chr.12: 31672299-31672299 on Build GRCh38
rs1003288985 Chr.12: 31672160-31672161 on Build GRCh38
rs1009607133 Chr.12: 31672264-31672264 on Build GRCh38
rs767926580 Chr.12: 31672345-31672345 on Build GRCh38
rs187003665 Chr.12: 31672193-31672193 on Build GRCh38
rs764751731 Chr.12: 31672289-31672289 on Build GRCh38
rs758636327 Chr.12: 31672283-31672283 on Build GRCh38
rs540084372 Chr.12: 31672244-31672244 on Build GRCh38
rs76114757 Chr.12: 31672326-31672326 on Build GRCh38
rs757706401 Chr.12: 31672332-31672332 on Build GRCh38
rs375155943 Chr.12: 31672300-31672300 on Build GRCh38
rs958505085 Chr.12: 31672313-31672313 on Build GRCh38
rs374483469 Chr.12: 31672259-31672259 on Build GRCh38
rs1053991252 Chr.12: 31672195-31672195 on Build GRCh38
rs751155970 Chr.12: 31672322-31672322 on Build GRCh38

View Full Product Details