Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 56745346-56745571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF3
Entrez Gene ID
50650
Gene Name
Rho guanine nucleotide exchange factor 3
Gene Aliases
GEF3, STA3, XPLN
UniGene
Hs.476402
Gene Chromosome Location
Chr.3: 56727418-57079308 on Build GRCh38
Transcripts
NM_001128615.1 NM_001128616.1 NM_001289698.1 NM_019555.2 XM_005265186.4 XM_005265187.3 XM_011533764.1 XM_011533765.1 XM_011533766.2 XM_011533767.1 XM_017006502.1 XM_017006503.1 XM_017006504.1 XM_017006505.1 XM_017006506.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7299897 c.764C>A p.A255D Chr.3: 56745407-56745407 on Build GRCh38
7299899 c.668C>A p.A223D Chr.3: 56745407-56745407 on Build GRCh38
3392527 c.624C>T p.L208L Chr.3: 56745451-56745451 on Build GRCh38
3993260 c.781A>C p.K261Q Chr.3: 56745390-56745390 on Build GRCh38
3993263 c.685A>C p.K229Q Chr.3: 56745390-56745390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776970187 Chr.3: 56745469-56745469 on Build GRCh38
rs550255987 Chr.3: 56745470-56745470 on Build GRCh38
rs34185780 Chr.3: 56745415-56745415 on Build GRCh38
rs1036110832 Chr.3: 56745476-56745476 on Build GRCh38
rs570138095 Chr.3: 56745505-56745505 on Build GRCh38
rs372602142 Chr.3: 56745377-56745377 on Build GRCh38
rs940814297 Chr.3: 56745532-56745532 on Build GRCh38
rs750007515 Chr.3: 56745504-56745504 on Build GRCh38
rs987708997 Chr.3: 56745517-56745517 on Build GRCh38
rs748328015 Chr.3: 56745409-56745409 on Build GRCh38
rs763258438 Chr.3: 56745510-56745510 on Build GRCh38
rs370046243 Chr.3: 56745481-56745481 on Build GRCh38
rs530670040 Chr.3: 56745467-56745467 on Build GRCh38
rs770236685 Chr.3: 56745475-56745475 on Build GRCh38
rs766954130 Chr.3: 56745495-56745495 on Build GRCh38
rs192861827 Chr.3: 56745521-56745521 on Build GRCh38
rs761493440 Chr.3: 56745490-56745490 on Build GRCh38
rs747042598 Chr.3: 56745444-56745444 on Build GRCh38
rs377224126 Chr.3: 56745414-56745414 on Build GRCh38

View Full Product Details