Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 37013783-37014048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD2
Entrez Gene ID
221472
Gene Name
FYVE, RhoGEF and PH domain containing 2
Gene Aliases
ZFYVE4
UniGene
Hs.509664
Gene Chromosome Location
Chr.6: 37005647-37029072 on Build GRCh38
Transcripts
NM_173558.3 XM_011514372.2 XM_011514373.2 XM_017010429.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3626316 c.752G>C p.R251P Chr.6: 37014029-37014029 on Build GRCh38
6663370 c.757G>A p.E253K Chr.6: 37014034-37014034 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761080837 Chr.6: 37013973-37013973 on Build GRCh38
rs757940226 Chr.6: 37014006-37014006 on Build GRCh38
rs374762712 Chr.6: 37013936-37013936 on Build GRCh38
rs754666163 Chr.6: 37013977-37013977 on Build GRCh38
rs867353455 Chr.6: 37014001-37014001 on Build GRCh38
rs536437800 Chr.6: 37013844-37013844 on Build GRCh38
rs774382924 Chr.6: 37013935-37013935 on Build GRCh38
rs149613118 Chr.6: 37013997-37013997 on Build GRCh38
rs759461969 Chr.6: 37013951-37013951 on Build GRCh38
rs576637403 Chr.6: 37013910-37013910 on Build GRCh38
rs773199799 Chr.6: 37013809-37013809 on Build GRCh38
rs987738505 Chr.6: 37013869-37013869 on Build GRCh38
rs1015956615 Chr.6: 37013856-37013856 on Build GRCh38
rs958404825 Chr.6: 37013823-37013823 on Build GRCh38
rs777897004 Chr.6: 37014013-37014013 on Build GRCh38
rs1013891137 Chr.6: 37013827-37013827 on Build GRCh38
rs145883518 Chr.6: 37013968-37013968 on Build GRCh38
rs768046681 Chr.6: 37013967-37013967 on Build GRCh38
rs774490665 Chr.6: 37013960-37013960 on Build GRCh38
rs1054856256 Chr.6: 37013924-37013924 on Build GRCh38
rs1023516227 Chr.6: 37013840-37013840 on Build GRCh38
rs760324468 Chr.6: 37014028-37014028 on Build GRCh38
rs201108665 Chr.6: 37013986-37013986 on Build GRCh38
rs377377669 Chr.6: 37013894-37013894 on Build GRCh38
rs999397386 Chr.6: 37013962-37013962 on Build GRCh38
rs144341931 Chr.6: 37014026-37014026 on Build GRCh38
rs867502871 Chr.6: 37013928-37013928 on Build GRCh38
rs754505925 Chr.6: 37013916-37013916 on Build GRCh38
rs372311354 Chr.6: 37013975-37013975 on Build GRCh38
rs908145734 Chr.6: 37013885-37013885 on Build GRCh38
rs761835247 Chr.6: 37013966-37013966 on Build GRCh38
rs763695122 Chr.6: 37014029-37014029 on Build GRCh38
rs143497870 Chr.6: 37013999-37013999 on Build GRCh38
rs746828814 Chr.6: 37013938-37013938 on Build GRCh38
rs756434090 Chr.6: 37013810-37013810 on Build GRCh38
rs940909221 Chr.6: 37013886-37013886 on Build GRCh38
rs369593356 Chr.6: 37013948-37013948 on Build GRCh38
rs367645913 Chr.6: 37013917-37013917 on Build GRCh38
rs374436730 Chr.6: 37013813-37013813 on Build GRCh38
rs34895385 Chr.6: 37013976-37013976 on Build GRCh38
rs567477782 Chr.6: 37013841-37013841 on Build GRCh38
rs193149955 Chr.6: 37013919-37013919 on Build GRCh38
rs753509955 Chr.6: 37013915-37013915 on Build GRCh38
rs1038309510 Chr.6: 37013889-37013889 on Build GRCh38
rs556760490 Chr.6: 37013868-37013868 on Build GRCh38
rs778373251 Chr.6: 37013987-37013987 on Build GRCh38
rs1013228592 Chr.6: 37013937-37013937 on Build GRCh38

View Full Product Details