Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23323928-23324155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL2L2-PABPN1
Entrez Gene ID
100529063
Gene Name
BCL2L2-PABPN1 readthrough
Gene Aliases
-
UniGene
Hs.707712
Gene Chromosome Location
Chr.14: 23306762-23326185 on Build GRCh38
Transcripts
NM_001199864.1 NM_004643.3

Gene Symbol
PABPN1
Entrez Gene ID
8106
Gene Name
poly(A) binding protein nuclear 1
Gene Aliases
OPMD, PAB2, PABII, PABP-2, PABP2
UniGene
Hs.707712
Gene Chromosome Location
Chr.14: 23320188-23326185 on Build GRCh38
Transcripts
NM_001199864.1 NM_004643.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1748760 c.662C>T p.S221L Chr.14: 23323985-23323985 on Build GRCh38
954673 c.648G>A p.A216A Chr.14: 23323971-23323971 on Build GRCh38
7215963 c.669A>G p.K223K Chr.14: 23323992-23323992 on Build GRCh38
4050005 c.720G>A p.R240R Chr.14: 23324043-23324043 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764111212 Chr.14: 23323995-23323995 on Build GRCh38
rs370676773 Chr.14: 23324016-23324016 on Build GRCh38
rs536320634 Chr.14: 23324084-23324084 on Build GRCh38
rs749698559 Chr.14: 23324060-23324060 on Build GRCh38
rs751852798 Chr.14: 23324086-23324086 on Build GRCh38
rs777004576 Chr.14: 23324066-23324066 on Build GRCh38
rs777404183 Chr.14: 23323980-23323980 on Build GRCh38
rs746611561 Chr.14: 23324065-23324065 on Build GRCh38
rs762127677 Chr.14: 23324067-23324067 on Build GRCh38
rs753693406 Chr.14: 23324095-23324095 on Build GRCh38
rs550945824 Chr.14: 23324059-23324059 on Build GRCh38
rs745799485 Chr.14: 23324106-23324106 on Build GRCh38
rs370788113 Chr.14: 23324071-23324071 on Build GRCh38
rs778930243 Chr.14: 23324004-23324004 on Build GRCh38
rs373643337 Chr.14: 23324031-23324031 on Build GRCh38
rs779409810 Chr.14: 23324115-23324115 on Build GRCh38
rs757288271 Chr.14: 23323992-23323992 on Build GRCh38
rs775283961 Chr.14: 23324061-23324061 on Build GRCh38
rs376401654 Chr.14: 23324038-23324038 on Build GRCh38
rs751724696 Chr.14: 23323970-23323970 on Build GRCh38
rs45528934 Chr.14: 23324096-23324096 on Build GRCh38
rs749040116 Chr.14: 23323988-23323988 on Build GRCh38
rs779056612 Chr.14: 23324103-23324103 on Build GRCh38
rs761855530 Chr.14: 23324107-23324107 on Build GRCh38
rs148847017 Chr.14: 23324013-23324013 on Build GRCh38
rs755974255 Chr.14: 23323971-23323971 on Build GRCh38
rs771474078 Chr.14: 23324058-23324058 on Build GRCh38
rs952249863 Chr.14: 23324109-23324109 on Build GRCh38
rs755175327 Chr.14: 23324093-23324093 on Build GRCh38
rs794727719 Chr.14: 23323984-23323984 on Build GRCh38
rs763159536 Chr.14: 23324081-23324081 on Build GRCh38
rs746481391 Chr.14: 23324117-23324117 on Build GRCh38
rs765745208 Chr.14: 23324078-23324078 on Build GRCh38

View Full Product Details