Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212047096-212047321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTL
Entrez Gene ID
51514
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Gene Aliases
CDT2, DCAF2, L2DTL, RAMP
UniGene
Hs.656473
Gene Chromosome Location
Chr.1: 212035553-212105006 on Build GRCh38
Transcripts
NM_001286229.1 NM_001286230.1 NM_016448.3 XM_011509614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6846157 c.289C>A p.H97N Chr.1: 212047162-212047162 on Build GRCh38
5422224 c.340-1G>T p.? Chr.1: 212047296-212047296 on Build GRCh38
5911301 c.323C>T p.P108L Chr.1: 212047196-212047196 on Build GRCh38
1930049 c.301G>A p.V101I Chr.1: 212047174-212047174 on Build GRCh38
6848197 c.331_332insTTAAACTTG p.V114_T115insKLV Chr.1: 212047204-212047205 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs865964377 Chr.1: 212047172-212047172 on Build GRCh38
rs578222368 Chr.1: 212047233-212047233 on Build GRCh38
rs771792975 Chr.1: 212047235-212047235 on Build GRCh38
rs758768497 Chr.1: 212047158-212047158 on Build GRCh38
rs749260131 Chr.1: 212047241-212047241 on Build GRCh38
rs186028431 Chr.1: 212047232-212047232 on Build GRCh38
rs371001369 Chr.1: 212047242-212047242 on Build GRCh38
rs537563413 Chr.1: 212047148-212047148 on Build GRCh38
rs1007844263 Chr.1: 212047282-212047282 on Build GRCh38
rs778216442 Chr.1: 212047159-212047159 on Build GRCh38
rs149991137 Chr.1: 212047138-212047138 on Build GRCh38
rs774233394 Chr.1: 212047248-212047248 on Build GRCh38
rs756969574 Chr.1: 212047174-212047174 on Build GRCh38
rs979484643 Chr.1: 212047131-212047131 on Build GRCh38
rs561544384 Chr.1: 212047126-212047126 on Build GRCh38
rs1056524471 Chr.1: 212047129-212047129 on Build GRCh38
rs754011833 Chr.1: 212047127-212047127 on Build GRCh38
rs780797655 Chr.1: 212047199-212047199 on Build GRCh38
rs79016895 Chr.1: 212047144-212047144 on Build GRCh38
rs751826465 Chr.1: 212047173-212047173 on Build GRCh38
rs746902125 Chr.1: 212047186-212047186 on Build GRCh38
rs745438451 Chr.1: 212047206-212047206 on Build GRCh38

View Full Product Details