Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6428243-6428502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPN
Entrez Gene ID
83715
Gene Name
espin
Gene Aliases
DFNB36, LP2654
UniGene
Hs.744222
Gene Chromosome Location
Chr.1: 6424776-6461367 on Build GRCh38
Transcripts
NM_031475.2 XM_005263501.2 XM_011542231.1 XM_011542232.1 XM_011542237.1 XM_011542238.2 XM_017002433.1 XM_017002434.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6064006 c.388G>A p.D130N Chr.1: 6428319-6428319 on Build GRCh38
7257636 c.398C>T p.A133V Chr.1: 6428329-6428329 on Build GRCh38
5723431 c.459C>T p.S153S Chr.1: 6428390-6428390 on Build GRCh38
7257450 c.390C>T p.D130D Chr.1: 6428321-6428321 on Build GRCh38
2238908 c.338G>A p.R113H Chr.1: 6428269-6428269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145666801 Chr.1: 6428313-6428313 on Build GRCh38
rs552379031 Chr.1: 6428312-6428312 on Build GRCh38
rs756669745 Chr.1: 6428346-6428346 on Build GRCh38
rs143543872 Chr.1: 6428338-6428338 on Build GRCh38
rs201813199 Chr.1: 6428273-6428273 on Build GRCh38
rs145002662 Chr.1: 6428403-6428403 on Build GRCh38
rs371554869 Chr.1: 6428451-6428451 on Build GRCh38
rs755009690 Chr.1: 6428310-6428310 on Build GRCh38
rs749289711 Chr.1: 6428369-6428369 on Build GRCh38
rs778970431 Chr.1: 6428410-6428410 on Build GRCh38
rs758026081 Chr.1: 6428279-6428279 on Build GRCh38
rs397833067 Chr.1: 6428406-6428406 on Build GRCh38
rs727504727 Chr.1: 6428370-6428370 on Build GRCh38
rs754914034 Chr.1: 6428373-6428373 on Build GRCh38
rs760314327 Chr.1: 6428441-6428441 on Build GRCh38
rs770342857 Chr.1: 6428393-6428393 on Build GRCh38
rs763679338 Chr.1: 6428282-6428282 on Build GRCh38
rs546577352 Chr.1: 6428442-6428442 on Build GRCh38
rs775996656 Chr.1: 6428402-6428402 on Build GRCh38
rs369494429 Chr.1: 6428444-6428444 on Build GRCh38
rs750010431 Chr.1: 6428353-6428353 on Build GRCh38
rs751233233 Chr.1: 6428283-6428283 on Build GRCh38
rs530495163 Chr.1: 6428281-6428281 on Build GRCh38
rs780647660 Chr.1: 6428310-6428310 on Build GRCh38
rs761424562 Chr.1: 6428411-6428411 on Build GRCh38
rs759977792 Chr.1: 6428337-6428337 on Build GRCh38
rs765867875 Chr.1: 6428274-6428274 on Build GRCh38
rs776191127 Chr.1: 6428339-6428339 on Build GRCh38
rs140382925 Chr.1: 6428344-6428344 on Build GRCh38
rs374134792 Chr.1: 6428330-6428330 on Build GRCh38
rs375866681 Chr.1: 6428461-6428461 on Build GRCh38
rs757407562 Chr.1: 6428460-6428460 on Build GRCh38
rs201911254 Chr.1: 6428405-6428405 on Build GRCh38
rs755951392 Chr.1: 6428358-6428358 on Build GRCh38
rs767227831 Chr.1: 6428348-6428348 on Build GRCh38
rs113326364 Chr.1: 6428398-6428398 on Build GRCh38
rs778659174 Chr.1: 6428379-6428379 on Build GRCh38
rs971782691 Chr.1: 6428475-6428475 on Build GRCh38
rs144234258 Chr.1: 6428363-6428363 on Build GRCh38
rs752557001 Chr.1: 6428448-6428449 on Build GRCh38
rs763542985 Chr.1: 6428340-6428340 on Build GRCh38
rs919425529 Chr.1: 6428323-6428323 on Build GRCh38
rs138979381 Chr.1: 6428328-6428328 on Build GRCh38
rs367832657 Chr.1: 6428269-6428269 on Build GRCh38
rs778941678 Chr.1: 6428448-6428448 on Build GRCh38
rs766911398 Chr.1: 6428431-6428431 on Build GRCh38
rs997445525 Chr.1: 6428397-6428397 on Build GRCh38
rs756932840 Chr.1: 6428306-6428306 on Build GRCh38
rs138239293 Chr.1: 6428327-6428327 on Build GRCh38
rs370070381 Chr.1: 6428445-6428445 on Build GRCh38
rs751089572 Chr.1: 6428345-6428345 on Build GRCh38
rs139223525 Chr.1: 6428382-6428382 on Build GRCh38
rs113467202 Chr.1: 6428329-6428329 on Build GRCh38
rs746069189 Chr.1: 6428387-6428387 on Build GRCh38
rs781518450 Chr.1: 6428384-6428384 on Build GRCh38
rs919425671 Chr.1: 6428479-6428479 on Build GRCh38
rs750146486 Chr.1: 6428436-6428436 on Build GRCh38
rs112712922 Chr.1: 6428321-6428321 on Build GRCh38
rs143577178 Chr.1: 6428268-6428268 on Build GRCh38

View Full Product Details