Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168907864-168908107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC2
Entrez Gene ID
57818
Gene Name
glucose-6-phosphatase catalytic subunit 2
Gene Aliases
IGRP
UniGene
Hs.283963
Gene Chromosome Location
Chr.2: 168901223-168910000 on Build GRCh38
Transcripts
NM_001081686.1 NM_021176.2 XM_011511564.2 XM_011511565.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3379851 c.861C>A p.N287K Chr.2: 168907872-168907872 on Build GRCh38
4989086 c.861C>G p.N287K Chr.2: 168907872-168907872 on Build GRCh38
6842944 c.910C>A p.L304M Chr.2: 168907921-168907921 on Build GRCh38
6036848 c.945C>T p.H315H Chr.2: 168907956-168907956 on Build GRCh38
6842939 c.873C>G p.S291R Chr.2: 168907884-168907884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541931958 Chr.2: 168907889-168907889 on Build GRCh38
rs201323443 Chr.2: 168907974-168907974 on Build GRCh38
rs754077966 Chr.2: 168908061-168908061 on Build GRCh38
rs772642710 Chr.2: 168907979-168907979 on Build GRCh38
rs757496063 Chr.2: 168908063-168908063 on Build GRCh38
rs769567058 Chr.2: 168908016-168908016 on Build GRCh38
rs925382715 Chr.2: 168907923-168907923 on Build GRCh38
rs141820044 Chr.2: 168907938-168907938 on Build GRCh38
rs201753573 Chr.2: 168907911-168907911 on Build GRCh38
rs745871593 Chr.2: 168907912-168907912 on Build GRCh38
rs779619992 Chr.2: 168907972-168907972 on Build GRCh38
rs373196602 Chr.2: 168907943-168907943 on Build GRCh38
rs760776009 Chr.2: 168908055-168908055 on Build GRCh38
rs2232328 Chr.2: 168908036-168908036 on Build GRCh38
rs777764289 Chr.2: 168907901-168907906 on Build GRCh38
rs11554658 Chr.2: 168908044-168908044 on Build GRCh38
rs574976550 Chr.2: 168907957-168907957 on Build GRCh38
rs777732226 Chr.2: 168908084-168908084 on Build GRCh38
rs923991987 Chr.2: 168908032-168908032 on Build GRCh38
rs768454510 Chr.2: 168908068-168908069 on Build GRCh38
rs760250729 Chr.2: 168907932-168907932 on Build GRCh38
rs765702651 Chr.2: 168908075-168908075 on Build GRCh38
rs370470317 Chr.2: 168907956-168907956 on Build GRCh38
rs549543851 Chr.2: 168907890-168907890 on Build GRCh38
rs768246460 Chr.2: 168908003-168908003 on Build GRCh38
rs137857125 Chr.2: 168907949-168907949 on Build GRCh38
rs200336133 Chr.2: 168907939-168907939 on Build GRCh38
rs1035992308 Chr.2: 168907892-168907892 on Build GRCh38
rs367851926 Chr.2: 168908060-168908060 on Build GRCh38
rs993287550 Chr.2: 168908046-168908046 on Build GRCh38
rs139331128 Chr.2: 168907929-168907929 on Build GRCh38
rs758253628 Chr.2: 168907979-168907979 on Build GRCh38
rs757689124 Chr.2: 168907971-168907971 on Build GRCh38
rs746746462 Chr.2: 168908067-168908067 on Build GRCh38
rs146233425 Chr.2: 168908000-168908000 on Build GRCh38
rs141041285 Chr.2: 168907913-168907913 on Build GRCh38
rs776320333 Chr.2: 168908005-168908005 on Build GRCh38
rs2232327 Chr.2: 168908030-168908030 on Build GRCh38
rs754193378 Chr.2: 168907960-168907960 on Build GRCh38
rs771693835 Chr.2: 168907917-168907917 on Build GRCh38
rs979502964 Chr.2: 168908014-168908014 on Build GRCh38
rs779531845 Chr.2: 168907893-168907893 on Build GRCh38
rs775478622 Chr.2: 168908051-168908051 on Build GRCh38
rs761665556 Chr.2: 168908013-168908013 on Build GRCh38
rs779945911 Chr.2: 168907991-168907997 on Build GRCh38
rs762906384 Chr.2: 168908041-168908041 on Build GRCh38
rs2232326 Chr.2: 168907981-168907981 on Build GRCh38
rs766251315 Chr.2: 168908043-168908043 on Build GRCh38
rs756530700 Chr.2: 168908073-168908073 on Build GRCh38
rs768200400 Chr.2: 168907933-168907933 on Build GRCh38

View Full Product Details