Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33280682-33280955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP5
Entrez Gene ID
51510
Gene Name
charged multivesicular body protein 5
Gene Aliases
C9orf83, CGI-34, HSPC177, PNAS-2, SNF7DC2, Vps60
UniGene
Hs.635313
Gene Chromosome Location
Chr.9: 33264879-33282069 on Build GRCh38
Transcripts
NM_001195536.1 NM_016410.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4152251 c.621G>C p.L207L Chr.9: 33280820-33280820 on Build GRCh38
1716474 c.658T>C p.*220Q Chr.9: 33280857-33280857 on Build GRCh38
6628490 c.657A>G p.S219S Chr.9: 33280856-33280856 on Build GRCh38
6628489 c.624G>A p.V208V Chr.9: 33280823-33280823 on Build GRCh38
3656913 c.610G>A p.D204N Chr.9: 33280809-33280809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35388632 Chr.9: 33280846-33280847 on Build GRCh38
rs776687504 Chr.9: 33280753-33280753 on Build GRCh38
rs201276553 Chr.9: 33280777-33280777 on Build GRCh38
rs543851609 Chr.9: 33280723-33280723 on Build GRCh38
rs79515112 Chr.9: 33280762-33280762 on Build GRCh38
rs759892223 Chr.9: 33280838-33280838 on Build GRCh38
rs745569191 Chr.9: 33280791-33280791 on Build GRCh38
rs757015643 Chr.9: 33280908-33280908 on Build GRCh38
rs55801336 Chr.9: 33280862-33280862 on Build GRCh38
rs1050157597 Chr.9: 33280820-33280820 on Build GRCh38
rs371931475 Chr.9: 33280870-33280870 on Build GRCh38
rs761084598 Chr.9: 33280823-33280823 on Build GRCh38
rs552765352 Chr.9: 33280847-33280847 on Build GRCh38
rs79502584 Chr.9: 33280763-33280763 on Build GRCh38
rs777521792 Chr.9: 33280889-33280889 on Build GRCh38
rs923826668 Chr.9: 33280737-33280737 on Build GRCh38
rs771469311 Chr.9: 33280730-33280730 on Build GRCh38
rs368264902 Chr.9: 33280804-33280804 on Build GRCh38
rs773880665 Chr.9: 33280816-33280816 on Build GRCh38
rs762799390 Chr.9: 33280892-33280894 on Build GRCh38
rs975719721 Chr.9: 33280752-33280752 on Build GRCh38
rs780781641 Chr.9: 33280780-33280780 on Build GRCh38
rs529642587 Chr.9: 33280778-33280778 on Build GRCh38
rs751528712 Chr.9: 33280897-33280897 on Build GRCh38
rs766894190 Chr.9: 33280832-33280832 on Build GRCh38
rs35307106 Chr.9: 33280776-33280777 on Build GRCh38
rs956625053 Chr.9: 33280738-33280738 on Build GRCh38
rs182128387 Chr.9: 33280805-33280805 on Build GRCh38
rs539912614 Chr.9: 33280785-33280785 on Build GRCh38
rs200709734 Chr.9: 33280814-33280814 on Build GRCh38
rs765351520 Chr.9: 33280839-33280839 on Build GRCh38
rs941504541 Chr.9: 33280872-33280872 on Build GRCh38
rs762531920 Chr.9: 33280810-33280810 on Build GRCh38
rs865967317 Chr.9: 33280807-33280807 on Build GRCh38

View Full Product Details