Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130930927-130931200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF4
Entrez Gene ID
50649
Gene Name
Rho guanine nucleotide exchange factor 4
Gene Aliases
ASEF, ASEF1, GEF4, STM6
UniGene
Hs.469935
Gene Chromosome Location
Chr.2: 130836916-131047253 on Build GRCh38
Transcripts
NM_015320.3 NM_032995.2 XM_005263681.1 XM_011511274.2 XM_017004231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5214914 c.55G>T p.A19S Chr.2: 130931012-130931012 on Build GRCh38
6087592 c.122G>C p.C41S Chr.2: 130931079-130931079 on Build GRCh38
5343719 c.123C>A p.C41* Chr.2: 130931080-130931080 on Build GRCh38
6604332 c.171C>A p.G57G Chr.2: 130931128-130931128 on Build GRCh38
441124 c.55G>T p.A19S Chr.2: 130931012-130931012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200806481 Chr.2: 130931102-130931102 on Build GRCh38
rs779181678 Chr.2: 130931136-130931136 on Build GRCh38
rs892129718 Chr.2: 130931019-130931019 on Build GRCh38
rs910776058 Chr.2: 130930967-130930967 on Build GRCh38
rs775063530 Chr.2: 130931013-130931013 on Build GRCh38
rs757812645 Chr.2: 130931172-130931172 on Build GRCh38
rs975036882 Chr.2: 130931128-130931128 on Build GRCh38
rs34035121 Chr.2: 130930967-130930968 on Build GRCh38
rs766179333 Chr.2: 130931041-130931041 on Build GRCh38
rs776243134 Chr.2: 130931108-130931108 on Build GRCh38
rs770999620 Chr.2: 130931026-130931026 on Build GRCh38
rs141778984 Chr.2: 130931147-130931147 on Build GRCh38
rs761262844 Chr.2: 130931021-130931021 on Build GRCh38
rs757648276 Chr.2: 130931089-130931089 on Build GRCh38
rs143550033 Chr.2: 130931146-130931146 on Build GRCh38
rs762720636 Chr.2: 130931040-130931040 on Build GRCh38
rs542503306 Chr.2: 130931057-130931057 on Build GRCh38
rs944987135 Chr.2: 130930988-130930988 on Build GRCh38
rs1055831425 Chr.2: 130931001-130931001 on Build GRCh38
rs373599530 Chr.2: 130931099-130931099 on Build GRCh38
rs199809047 Chr.2: 130931022-130931022 on Build GRCh38
rs10188052 Chr.2: 130931054-130931054 on Build GRCh38
rs778492515 Chr.2: 130931072-130931072 on Build GRCh38
rs6718816 Chr.2: 130931014-130931014 on Build GRCh38
rs778404872 Chr.2: 130930958-130930958 on Build GRCh38
rs745911648 Chr.2: 130931171-130931172 on Build GRCh38
rs776148463 Chr.2: 130931020-130931020 on Build GRCh38
rs752965067 Chr.2: 130931159-130931159 on Build GRCh38
rs541543130 Chr.2: 130930957-130930957 on Build GRCh38
rs771323502 Chr.2: 130930992-130930992 on Build GRCh38
rs35359593 Chr.2: 130931127-130931127 on Build GRCh38
rs756233817 Chr.2: 130931069-130931069 on Build GRCh38
rs367660031 Chr.2: 130931138-130931138 on Build GRCh38
rs375855121 Chr.2: 130931090-130931090 on Build GRCh38
rs150071332 Chr.2: 130930965-130930965 on Build GRCh38
rs749789415 Chr.2: 130931074-130931074 on Build GRCh38
rs753795389 Chr.2: 130931171-130931171 on Build GRCh38
rs767803461 Chr.2: 130931058-130931058 on Build GRCh38
rs762957799 Chr.2: 130931139-130931139 on Build GRCh38
rs756321975 Chr.2: 130931165-130931165 on Build GRCh38
rs774227641 Chr.2: 130931143-130931143 on Build GRCh38
rs527734596 Chr.2: 130931100-130931100 on Build GRCh38
rs914839675 Chr.2: 130931173-130931173 on Build GRCh38
rs766190445 Chr.2: 130931142-130931142 on Build GRCh38
rs764282836 Chr.2: 130931166-130931166 on Build GRCh38
rs747693778 Chr.2: 130931127-130931127 on Build GRCh38
rs773167697 Chr.2: 130931030-130931030 on Build GRCh38
rs549714029 Chr.2: 130931124-130931124 on Build GRCh38
rs202134576 Chr.2: 130930947-130930947 on Build GRCh38
rs34254937 Chr.2: 130931041-130931042 on Build GRCh38

View Full Product Details