Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18912869-18913120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPE
Entrez Gene ID
11316
Gene Name
coatomer protein complex subunit epsilon
Gene Aliases
epsilon-COP
UniGene
Hs.10326
Gene Chromosome Location
Chr.19: 18899514-18919464 on Build GRCh38
Transcripts
NM_007263.3 NM_199442.1 NM_199444.1 XM_011527655.1 XM_011527656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634052 c.177G>A p.A59A Chr.19: 18912996-18912996 on Build GRCh38
1238578 c.160G>A p.V54I Chr.19: 18913013-18913013 on Build GRCh38
1665425 c.145_146delGA p.D49fs*9 Chr.19: 18913027-18913028 on Build GRCh38
7280393 c.189+8C>T p.? Chr.19: 18912976-18912976 on Build GRCh38
6634050 c.176C>T p.A59V Chr.19: 18912997-18912997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528549603 Chr.19: 18913079-18913079 on Build GRCh38
rs930169839 Chr.19: 18912915-18912918 on Build GRCh38
rs944179536 Chr.19: 18913011-18913011 on Build GRCh38
rs111654958 Chr.19: 18913072-18913072 on Build GRCh38
rs200759503 Chr.19: 18913029-18913029 on Build GRCh38
rs748323022 Chr.19: 18912977-18912977 on Build GRCh38
rs765310706 Chr.19: 18912973-18912974 on Build GRCh38
rs750889117 Chr.19: 18913014-18913014 on Build GRCh38
rs985709633 Chr.19: 18912994-18912994 on Build GRCh38
rs563566069 Chr.19: 18913020-18913020 on Build GRCh38
rs140598767 Chr.19: 18913003-18913003 on Build GRCh38
rs751145190 Chr.19: 18912939-18912939 on Build GRCh38
rs777738643 Chr.19: 18913053-18913053 on Build GRCh38
rs768265562 Chr.19: 18913065-18913065 on Build GRCh38
rs755241399 Chr.19: 18913026-18913026 on Build GRCh38
rs200131883 Chr.19: 18913025-18913025 on Build GRCh38
rs769789174 Chr.19: 18913059-18913059 on Build GRCh38
rs191926085 Chr.19: 18913082-18913082 on Build GRCh38
rs749460373 Chr.19: 18912965-18912965 on Build GRCh38
rs752689594 Chr.19: 18913032-18913032 on Build GRCh38
rs768353462 Chr.19: 18912990-18912990 on Build GRCh38
rs761390501 Chr.19: 18913069-18913069 on Build GRCh38
rs761274738 Chr.19: 18913018-18913018 on Build GRCh38
rs759542266 Chr.19: 18912938-18912938 on Build GRCh38
rs745748897 Chr.19: 18913058-18913058 on Build GRCh38
rs749130175 Chr.19: 18913061-18913061 on Build GRCh38
rs779132915 Chr.19: 18912976-18912976 on Build GRCh38
rs765552123 Chr.19: 18913092-18913092 on Build GRCh38
rs942944007 Chr.19: 18912950-18912950 on Build GRCh38
rs755259484 Chr.19: 18912975-18912975 on Build GRCh38
rs371028688 Chr.19: 18912996-18912996 on Build GRCh38
rs368396421 Chr.19: 18912979-18912979 on Build GRCh38
rs912701226 Chr.19: 18912947-18912947 on Build GRCh38
rs761262388 Chr.19: 18913078-18913078 on Build GRCh38
rs367852757 Chr.19: 18913091-18913091 on Build GRCh38
rs754322650 Chr.19: 18913024-18913024 on Build GRCh38
rs3177137 Chr.19: 18913044-18913044 on Build GRCh38
rs762085966 Chr.19: 18912988-18912988 on Build GRCh38
rs374487488 Chr.19: 18912964-18912964 on Build GRCh38
rs112377830 Chr.19: 18913049-18913049 on Build GRCh38
rs371020325 Chr.19: 18913013-18913013 on Build GRCh38
rs992263390 Chr.19: 18912902-18912902 on Build GRCh38
rs757691454 Chr.19: 18912949-18912949 on Build GRCh38
rs768117439 Chr.19: 18912954-18912954 on Build GRCh38
rs774011313 Chr.19: 18913068-18913068 on Build GRCh38
rs779809889 Chr.19: 18913060-18913060 on Build GRCh38
rs745824851 Chr.19: 18912984-18912984 on Build GRCh38
rs560580883 Chr.19: 18913087-18913087 on Build GRCh38
rs929738905 Chr.19: 18913045-18913045 on Build GRCh38
rs144870827 Chr.19: 18912987-18912987 on Build GRCh38
rs1051069081 Chr.19: 18913063-18913063 on Build GRCh38
rs756502021 Chr.19: 18912961-18912961 on Build GRCh38

View Full Product Details