Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38326849-38327112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR179
Entrez Gene ID
440435
Gene Name
G protein-coupled receptor 179
Gene Aliases
CSNB1E, GPR158L, GPR158L1
UniGene
Hs.462915
Gene Chromosome Location
Chr.17: 38325610-38343810 on Build GRCh38
Transcripts
NM_001004334.3

SNP Detail

SNP ID Chromosome Location
rs749176781 Chr.17: 38326961-38326961 on Build GRCh38
rs574735535 Chr.17: 38326937-38326937 on Build GRCh38
rs762181891 Chr.17: 38326955-38326956 on Build GRCh38
rs770150645 Chr.17: 38327003-38327036 on Build GRCh38
rs757224017 Chr.17: 38326938-38326938 on Build GRCh38
rs1003419531 Chr.17: 38326922-38326922 on Build GRCh38
rs879199706 Chr.17: 38326960-38326960 on Build GRCh38
rs370368728 Chr.17: 38326883-38326883 on Build GRCh38
rs374054393 Chr.17: 38326886-38326886 on Build GRCh38
rs899602839 Chr.17: 38326980-38326980 on Build GRCh38
rs199518600 Chr.17: 38327039-38327039 on Build GRCh38
rs201755354 Chr.17: 38326904-38326904 on Build GRCh38
rs368811282 Chr.17: 38327048-38327048 on Build GRCh38
rs765920613 Chr.17: 38326878-38326878 on Build GRCh38
rs772935790 Chr.17: 38327054-38327054 on Build GRCh38
rs778825639 Chr.17: 38326984-38326984 on Build GRCh38
rs1048141576 Chr.17: 38327070-38327070 on Build GRCh38
rs781391422 Chr.17: 38326948-38326948 on Build GRCh38
rs368870706 Chr.17: 38327079-38327079 on Build GRCh38
rs200763394 Chr.17: 38327073-38327073 on Build GRCh38
rs114844749 Chr.17: 38327075-38327075 on Build GRCh38
rs540513728 Chr.17: 38326985-38326985 on Build GRCh38
rs368207971 Chr.17: 38326900-38326900 on Build GRCh38
rs535678195 Chr.17: 38326932-38326932 on Build GRCh38
rs201374947 Chr.17: 38327074-38327074 on Build GRCh38
rs765784093 Chr.17: 38327076-38327076 on Build GRCh38
rs190716378 Chr.17: 38326928-38326928 on Build GRCh38
rs768675419 Chr.17: 38326977-38326977 on Build GRCh38
rs555500617 Chr.17: 38327037-38327037 on Build GRCh38

View Full Product Details