Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 1746801-1747054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN1
Entrez Gene ID
392617
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
PPP1R28
UniGene
Hs.42896
Gene Chromosome Location
Chr.7: 1666075-1747954 on Build GRCh38
Transcripts
NM_001128636.2 XM_006715725.3 XM_006715726.2 XM_006715727.3 XM_011515397.2 XM_011515398.2 XM_011515401.2 XM_017012203.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4565659 c.2364_2365GG>AA p.E789K Chr.7: 1746960-1746961 on Build GRCh38
5544898 c.2365G>A p.E789K Chr.7: 1746961-1746961 on Build GRCh38
3411880 c.2416T>C p.F806L Chr.7: 1747012-1747012 on Build GRCh38
3394652 c.2353C>T p.R785W Chr.7: 1746949-1746949 on Build GRCh38
5807343 c.2225G>A p.R742Q Chr.7: 1746821-1746821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781221517 Chr.7: 1746838-1746838 on Build GRCh38
rs897812110 Chr.7: 1746946-1746946 on Build GRCh38
rs757469332 Chr.7: 1746985-1746985 on Build GRCh38
rs868205051 Chr.7: 1746961-1746961 on Build GRCh38
rs777702721 Chr.7: 1747034-1747034 on Build GRCh38
rs933268572 Chr.7: 1746833-1746833 on Build GRCh38
rs748145478 Chr.7: 1746901-1746901 on Build GRCh38
rs554133089 Chr.7: 1746841-1746841 on Build GRCh38
rs773186780 Chr.7: 1746931-1746931 on Build GRCh38
rs907438302 Chr.7: 1746920-1746920 on Build GRCh38
rs371301012 Chr.7: 1746845-1746845 on Build GRCh38
rs773976889 Chr.7: 1746821-1746821 on Build GRCh38
rs374274179 Chr.7: 1746848-1746848 on Build GRCh38
rs543465852 Chr.7: 1746895-1746895 on Build GRCh38
rs1020517722 Chr.7: 1746908-1746908 on Build GRCh38
rs574238696 Chr.7: 1746894-1746894 on Build GRCh38
rs889826376 Chr.7: 1746840-1746840 on Build GRCh38
rs942793771 Chr.7: 1746853-1746853 on Build GRCh38
rs1010838225 Chr.7: 1746902-1746902 on Build GRCh38
rs757525002 Chr.7: 1746832-1746832 on Build GRCh38
rs758397436 Chr.7: 1747023-1747023 on Build GRCh38
rs923131587 Chr.7: 1747018-1747018 on Build GRCh38
rs770773878 Chr.7: 1746948-1746948 on Build GRCh38
rs756084251 Chr.7: 1747001-1747001 on Build GRCh38
rs971984304 Chr.7: 1746825-1746825 on Build GRCh38
rs1054957619 Chr.7: 1746859-1746859 on Build GRCh38
rs937935582 Chr.7: 1746834-1746834 on Build GRCh38
rs771713224 Chr.7: 1746924-1746924 on Build GRCh38
rs577340198 Chr.7: 1746984-1746984 on Build GRCh38
rs750674255 Chr.7: 1746990-1746990 on Build GRCh38
rs111457141 Chr.7: 1746953-1746953 on Build GRCh38
rs781059252 Chr.7: 1746965-1746966 on Build GRCh38
rs775822166 Chr.7: 1746970-1746970 on Build GRCh38
rs200235323 Chr.7: 1746987-1746987 on Build GRCh38
rs746626090 Chr.7: 1746947-1746947 on Build GRCh38
rs754932894 Chr.7: 1746844-1746844 on Build GRCh38
rs748011357 Chr.7: 1747014-1747014 on Build GRCh38
rs557288458 Chr.7: 1746952-1746952 on Build GRCh38
rs559728199 Chr.7: 1746864-1746864 on Build GRCh38
rs370334516 Chr.7: 1746981-1746981 on Build GRCh38
rs1036475500 Chr.7: 1746932-1746932 on Build GRCh38
rs751731833 Chr.7: 1746827-1746827 on Build GRCh38
rs776267027 Chr.7: 1746949-1746949 on Build GRCh38
rs958649118 Chr.7: 1746967-1746967 on Build GRCh38
rs780202361 Chr.7: 1747010-1747010 on Build GRCh38
rs764807037 Chr.7: 1746968-1746968 on Build GRCh38

View Full Product Details