Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 11617478-11617703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
UniGene
Hs.567259
Gene Chromosome Location
Chr.17: 11598431-11970168 on Build GRCh38
Transcripts
NM_001372.3 XM_011523703.1 XM_017024292.1 XM_017024294.1 XM_017024295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3818817 c.1105C>G p.L369V Chr.17: 11617611-11617611 on Build GRCh38
1731023 c.1071G>T p.R357S Chr.17: 11617577-11617577 on Build GRCh38
5041031 c.1085T>C p.L362P Chr.17: 11617591-11617591 on Build GRCh38
344478 c.990G>T p.E330D Chr.17: 11617496-11617496 on Build GRCh38
4063912 c.1008G>A p.R336R Chr.17: 11617514-11617514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777169126 Chr.17: 11617550-11617550 on Build GRCh38
rs368678135 Chr.17: 11617568-11617568 on Build GRCh38
rs747356088 Chr.17: 11617546-11617546 on Build GRCh38
rs753260194 Chr.17: 11617572-11617572 on Build GRCh38
rs748588408 Chr.17: 11617612-11617612 on Build GRCh38
rs1002730051 Chr.17: 11617527-11617527 on Build GRCh38
rs778588804 Chr.17: 11617577-11617577 on Build GRCh38
rs954888924 Chr.17: 11617499-11617499 on Build GRCh38
rs781679184 Chr.17: 11617596-11617596 on Build GRCh38
rs771418052 Chr.17: 11617564-11617564 on Build GRCh38
rs745479283 Chr.17: 11617584-11617584 on Build GRCh38
rs746319400 Chr.17: 11617641-11617641 on Build GRCh38
rs747927532 Chr.17: 11617566-11617567 on Build GRCh38
rs778670254 Chr.17: 11617540-11617540 on Build GRCh38
rs151151666 Chr.17: 11617556-11617556 on Build GRCh38
rs576365392 Chr.17: 11617616-11617616 on Build GRCh38
rs1052916090 Chr.17: 11617583-11617583 on Build GRCh38
rs766487288 Chr.17: 11617566-11617566 on Build GRCh38
rs369392784 Chr.17: 11617526-11617526 on Build GRCh38
rs770389199 Chr.17: 11617557-11617557 on Build GRCh38
rs928524930 Chr.17: 11617581-11617581 on Build GRCh38
rs758130208 Chr.17: 11617593-11617593 on Build GRCh38
rs759693117 Chr.17: 11617662-11617662 on Build GRCh38
rs756842427 Chr.17: 11617573-11617573 on Build GRCh38
rs373491778 Chr.17: 11617570-11617570 on Build GRCh38
rs79601463 Chr.17: 11617658-11617658 on Build GRCh38
rs183304006 Chr.17: 11617630-11617630 on Build GRCh38
rs769093335 Chr.17: 11617547-11617547 on Build GRCh38
rs767318637 Chr.17: 11617503-11617503 on Build GRCh38
rs199639850 Chr.17: 11617512-11617512 on Build GRCh38
rs767788610 Chr.17: 11617670-11617670 on Build GRCh38
rs938534383 Chr.17: 11617582-11617582 on Build GRCh38
rs572251457 Chr.17: 11617569-11617569 on Build GRCh38
rs554701984 Chr.17: 11617571-11617571 on Build GRCh38
rs886968718 Chr.17: 11617511-11617511 on Build GRCh38
rs377415601 Chr.17: 11617563-11617563 on Build GRCh38
rs995830083 Chr.17: 11617660-11617660 on Build GRCh38
rs748889510 Chr.17: 11617521-11617521 on Build GRCh38
rs565544220 Chr.17: 11617642-11617642 on Build GRCh38
rs756079633 Chr.17: 11617513-11617513 on Build GRCh38
rs3744574 Chr.17: 11617516-11617516 on Build GRCh38
rs891549283 Chr.17: 11617586-11617586 on Build GRCh38
rs1035905911 Chr.17: 11617541-11617541 on Build GRCh38
rs973887639 Chr.17: 11617559-11617559 on Build GRCh38

View Full Product Details