Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 120982685-120982954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLI2
Entrez Gene ID
2736
Gene Name
GLI family zinc finger 2
Gene Aliases
CJS, HPE9, PHS2, THP1, THP2
UniGene
Hs.111867
Gene Chromosome Location
Chr.2: 120797291-120992653 on Build GRCh38
Transcripts
NM_005270.4 XM_006712422.2 XM_011510969.2 XM_011510970.2 XM_011510971.2 XM_011510972.2 XM_011510973.2 XM_011510974.2 XM_017003818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3836702 c.1529G>A p.C510Y Chr.2: 120982726-120982726 on Build GRCh38
5676111 c.1619G>C p.C540S Chr.2: 120982816-120982816 on Build GRCh38
1613347 c.1560G>A p.L520L Chr.2: 120982757-120982757 on Build GRCh38
6671503 c.1612G>A p.E538K Chr.2: 120982809-120982809 on Build GRCh38
3565904 c.1544C>T p.S515F Chr.2: 120982741-120982741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772471844 Chr.2: 120982878-120982878 on Build GRCh38
rs766011846 Chr.2: 120982799-120982799 on Build GRCh38
rs896259039 Chr.2: 120982809-120982809 on Build GRCh38
rs372925840 Chr.2: 120982729-120982729 on Build GRCh38
rs781146798 Chr.2: 120982719-120982719 on Build GRCh38
rs775943788 Chr.2: 120982758-120982758 on Build GRCh38
rs747520338 Chr.2: 120982884-120982884 on Build GRCh38
rs280196 Chr.2: 120982929-120982929 on Build GRCh38
rs762489303 Chr.2: 120982899-120982899 on Build GRCh38
rs1008569568 Chr.2: 120982707-120982707 on Build GRCh38
rs762529311 Chr.2: 120982787-120982787 on Build GRCh38
rs117209625 Chr.2: 120982916-120982916 on Build GRCh38
rs551585465 Chr.2: 120982912-120982912 on Build GRCh38
rs746195258 Chr.2: 120982743-120982743 on Build GRCh38
rs752601850 Chr.2: 120982892-120982902 on Build GRCh38
rs746257608 Chr.2: 120982877-120982877 on Build GRCh38
rs983304510 Chr.2: 120982865-120982865 on Build GRCh38
rs267598855 Chr.2: 120982824-120982824 on Build GRCh38
rs564753456 Chr.2: 120982845-120982845 on Build GRCh38
rs200621415 Chr.2: 120982790-120982790 on Build GRCh38
rs780145025 Chr.2: 120982709-120982711 on Build GRCh38
rs369337841 Chr.2: 120982930-120982930 on Build GRCh38
rs772643710 Chr.2: 120982782-120982782 on Build GRCh38
rs772735571 Chr.2: 120982896-120982896 on Build GRCh38
rs542656225 Chr.2: 120982745-120982745 on Build GRCh38
rs137960978 Chr.2: 120982781-120982781 on Build GRCh38
rs749322649 Chr.2: 120982847-120982847 on Build GRCh38
rs759304264 Chr.2: 120982913-120982913 on Build GRCh38
rs779153592 Chr.2: 120982874-120982874 on Build GRCh38
rs761373106 Chr.2: 120982778-120982778 on Build GRCh38
rs774591307 Chr.2: 120982734-120982734 on Build GRCh38
rs544791528 Chr.2: 120982718-120982718 on Build GRCh38
rs200300186 Chr.2: 120982841-120982841 on Build GRCh38
rs368332990 Chr.2: 120982704-120982704 on Build GRCh38
rs1000519306 Chr.2: 120982910-120982910 on Build GRCh38
rs767400136 Chr.2: 120982835-120982835 on Build GRCh38
rs201279367 Chr.2: 120982825-120982825 on Build GRCh38
rs769094105 Chr.2: 120982890-120982890 on Build GRCh38
rs200507501 Chr.2: 120982730-120982730 on Build GRCh38
rs773572118 Chr.2: 120982868-120982868 on Build GRCh38
rs140347335 Chr.2: 120982891-120982891 on Build GRCh38
rs546968102 Chr.2: 120982933-120982933 on Build GRCh38
rs751314552 Chr.2: 120982713-120982713 on Build GRCh38
rs41279790 Chr.2: 120982917-120982917 on Build GRCh38
rs765991375 Chr.2: 120982909-120982909 on Build GRCh38
rs756237921 Chr.2: 120982722-120982722 on Build GRCh38
rs753895640 Chr.2: 120982920-120982920 on Build GRCh38

View Full Product Details