Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48742550-48742823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL4
Entrez Gene ID
84871
Gene Name
ATP/GTP binding protein like 4
Gene Aliases
CCP6
UniGene
Hs.551844
Gene Chromosome Location
Chr.1: 48520157-50024011 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1 NM_001302082.1 NM_024603.3 XM_011542141.2 XM_011542142.2 XM_017002331.1 XM_017002332.1 XM_017002333.1 XM_017002334.1

Gene Symbol
BEND5
Entrez Gene ID
79656
Gene Name
BEN domain containing 5
Gene Aliases
C1orf165
UniGene
Hs.475348
Gene Chromosome Location
Chr.1: 48727523-48778603 on Build GRCh38
Transcripts
NM_001323573.1 NM_001323574.1 NM_001323575.1 NM_032785.3 NR_136623.1 XM_011542308.2 XM_017002595.1 XM_017002596.1 XM_017002597.1 NM_001302082.1 NM_024603.3 XM_011542141.2 XM_011542142.2 XM_017002331.1 XM_017002332.1 XM_017002333.1 XM_017002334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6611271 c.850G>A p.A284T Chr.1: 48742667-48742667 on Build GRCh38
2192017 c.836C>T p.S279L Chr.1: 48742681-48742681 on Build GRCh38
2192018 c.329C>T p.S110L Chr.1: 48742681-48742681 on Build GRCh38
6388211 c.361G>T p.D121Y Chr.1: 48742649-48742649 on Build GRCh38
1184659 c.806G>T p.R269L Chr.1: 48742711-48742711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149339808 Chr.1: 48742644-48742644 on Build GRCh38
rs964008968 Chr.1: 48742712-48742712 on Build GRCh38
rs200465195 Chr.1: 48742773-48742773 on Build GRCh38
rs762724446 Chr.1: 48742591-48742591 on Build GRCh38
rs750400862 Chr.1: 48742647-48742647 on Build GRCh38
rs747568526 Chr.1: 48742675-48742675 on Build GRCh38
rs756160114 Chr.1: 48742652-48742652 on Build GRCh38
rs761563513 Chr.1: 48742720-48742720 on Build GRCh38
rs757862667 Chr.1: 48742574-48742577 on Build GRCh38
rs143430976 Chr.1: 48742683-48742683 on Build GRCh38
rs771957986 Chr.1: 48742743-48742743 on Build GRCh38
rs374644523 Chr.1: 48742666-48742666 on Build GRCh38
rs766306124 Chr.1: 48742731-48742731 on Build GRCh38
rs781065724 Chr.1: 48742645-48742645 on Build GRCh38
rs550048480 Chr.1: 48742769-48742769 on Build GRCh38
rs34838490 Chr.1: 48742680-48742680 on Build GRCh38
rs747515236 Chr.1: 48742775-48742775 on Build GRCh38
rs902044046 Chr.1: 48742711-48742711 on Build GRCh38
rs779159389 Chr.1: 48742749-48742749 on Build GRCh38
rs756106934 Chr.1: 48742730-48742730 on Build GRCh38
rs996357621 Chr.1: 48742714-48742714 on Build GRCh38
rs146041042 Chr.1: 48742668-48742668 on Build GRCh38
rs748499198 Chr.1: 48742667-48742667 on Build GRCh38
rs752270772 Chr.1: 48742576-48742576 on Build GRCh38
rs769416308 Chr.1: 48742677-48742677 on Build GRCh38
rs144681129 Chr.1: 48742659-48742659 on Build GRCh38
rs1029794726 Chr.1: 48742746-48742746 on Build GRCh38
rs200112741 Chr.1: 48742724-48742724 on Build GRCh38
rs371459284 Chr.1: 48742613-48742613 on Build GRCh38
rs369662437 Chr.1: 48742757-48742757 on Build GRCh38
rs755038158 Chr.1: 48742747-48742747 on Build GRCh38
rs754514536 Chr.1: 48742663-48742667 on Build GRCh38
rs527961194 Chr.1: 48742605-48742605 on Build GRCh38
rs779505632 Chr.1: 48742580-48742583 on Build GRCh38
rs780162989 Chr.1: 48742657-48742657 on Build GRCh38
rs753895119 Chr.1: 48742745-48742745 on Build GRCh38
rs763751863 Chr.1: 48742701-48742701 on Build GRCh38
rs755159747 Chr.1: 48742660-48742660 on Build GRCh38
rs780743938 Chr.1: 48742673-48742674 on Build GRCh38
rs771499627 Chr.1: 48742781-48742781 on Build GRCh38
rs377372975 Chr.1: 48742725-48742725 on Build GRCh38
rs773905436 Chr.1: 48742706-48742706 on Build GRCh38
rs148772068 Chr.1: 48742767-48742767 on Build GRCh38
rs767956340 Chr.1: 48742594-48742594 on Build GRCh38
rs747945703 Chr.1: 48742662-48742662 on Build GRCh38

View Full Product Details