Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 61817338-61817569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH8
Entrez Gene ID
1006
Gene Name
cadherin 8
Gene Aliases
Nbla04261
UniGene
Hs.368322
Gene Chromosome Location
Chr.16: 61640435-62036835 on Build GRCh38
Transcripts
NM_001796.4 XM_005255760.3 XM_017022829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
703966 c.1216C>G p.L406V Chr.16: 61817540-61817540 on Build GRCh38
4989411 c.1210G>C p.A404P Chr.16: 61817546-61817546 on Build GRCh38
6036530 c.1247G>T p.R416L Chr.16: 61817509-61817509 on Build GRCh38
6233533 c.1270C>T p.P424S Chr.16: 61817486-61817486 on Build GRCh38
5013022 c.1273A>T p.I425L Chr.16: 61817483-61817483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774332279 Chr.16: 61817485-61817485 on Build GRCh38
rs779227539 Chr.16: 61817526-61817526 on Build GRCh38
rs944025686 Chr.16: 61817377-61817377 on Build GRCh38
rs151208133 Chr.16: 61817509-61817509 on Build GRCh38
rs142168278 Chr.16: 61817439-61817439 on Build GRCh38
rs908817456 Chr.16: 61817369-61817369 on Build GRCh38
rs780237284 Chr.16: 61817531-61817531 on Build GRCh38
rs767418229 Chr.16: 61817490-61817490 on Build GRCh38
rs918661300 Chr.16: 61817394-61817394 on Build GRCh38
rs768590514 Chr.16: 61817473-61817473 on Build GRCh38
rs760119610 Chr.16: 61817502-61817502 on Build GRCh38
rs367989208 Chr.16: 61817496-61817496 on Build GRCh38
rs758903938 Chr.16: 61817539-61817539 on Build GRCh38
rs1047189836 Chr.16: 61817359-61817361 on Build GRCh38
rs374093874 Chr.16: 61817452-61817452 on Build GRCh38
rs887576259 Chr.16: 61817412-61817412 on Build GRCh38
rs762794934 Chr.16: 61817466-61817466 on Build GRCh38
rs375849945 Chr.16: 61817540-61817540 on Build GRCh38
rs772619039 Chr.16: 61817437-61817437 on Build GRCh38
rs73564352 Chr.16: 61817410-61817410 on Build GRCh38
rs976893175 Chr.16: 61817382-61817382 on Build GRCh38
rs747114374 Chr.16: 61817451-61817451 on Build GRCh38
rs531546217 Chr.16: 61817532-61817532 on Build GRCh38
rs569050495 Chr.16: 61817523-61817523 on Build GRCh38
rs149155669 Chr.16: 61817405-61817405 on Build GRCh38
rs1030798995 Chr.16: 61817541-61817541 on Build GRCh38
rs375943951 Chr.16: 61817431-61817431 on Build GRCh38
rs938896925 Chr.16: 61817398-61817398 on Build GRCh38
rs761808168 Chr.16: 61817489-61817489 on Build GRCh38
rs911242272 Chr.16: 61817373-61817373 on Build GRCh38
rs548909736 Chr.16: 61817510-61817510 on Build GRCh38
rs532834411 Chr.16: 61817429-61817429 on Build GRCh38
rs894841480 Chr.16: 61817456-61817456 on Build GRCh38
rs765994153 Chr.16: 61817505-61817505 on Build GRCh38

View Full Product Details