Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 143951863-143952136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTDC1
Entrez Gene ID
79712
Gene Name
glycosyltransferase like domain containing 1
Gene Aliases
Hmat-Xa, mat-Xa
UniGene
Hs.44780
Gene Chromosome Location
Chr.2: 143945787-144333092 on Build GRCh38
Transcripts
NM_001006636.3 NM_001164629.3 NM_001284233.1 NM_001284234.1 NM_001284238.1 NM_024659.4 XM_005263774.2 XM_005263782.2 XM_005263784.2 XM_011511843.2 XM_011511855.2 XM_011511856.2 XM_017004917.1 XM_017004918.1 XM_017004919.1 XM_017004920.1 XM_017004921.1 XM_017004922.1 XM_017004923.1 XM_017004924.1 XM_017004925.1 XM_017004926.1 XM_017004927.1 XM_017004928.1 XM_017004929.1 XM_017004930.1 XM_017004931.1 XM_017004932.1 XM_017004933.1 XM_017004934.1 XM_017004935.1 XM_017004936.1 XM_017004937.1 XM_017004938.1 XM_017004939.1 XM_017004940.1 XM_017004941.1 XM_017004942.1 XM_017004943.1 XM_017004944.1 XM_017004945.1 XM_017004946.1 XM_017004947.1 XM_017004948.1 XM_017004949.1 XM_017004950.1 XM_017004951.1 XM_017004952.1 XM_017004953.1 XM_017004954.1 XM_017004955.1 XM_017004956.1 XM_017004957.1 NR_110237.1

Gene Symbol
LOC101928386
Entrez Gene ID
101928386
Gene Name
uncharacterized LOC101928386
Gene Aliases
-
UniGene
Hs.668251
Gene Chromosome Location
Chr.2: 143937067-143964156 on Build GRCh38
Transcripts
NM_001006636.3 NM_001164629.3 NM_001284233.1 NM_001284234.1 NM_001284238.1 NM_024659.4 XM_005263774.2 XM_005263782.2 XM_005263784.2 XM_011511843.2 XM_011511855.2 XM_011511856.2 XM_017004917.1 XM_017004918.1 XM_017004919.1 XM_017004920.1 XM_017004921.1 XM_017004922.1 XM_017004923.1 XM_017004924.1 XM_017004925.1 XM_017004926.1 XM_017004927.1 XM_017004928.1 XM_017004929.1 XM_017004930.1 XM_017004931.1 XM_017004932.1 XM_017004933.1 XM_017004934.1 XM_017004935.1 XM_017004936.1 XM_017004937.1 XM_017004938.1 XM_017004939.1 XM_017004940.1 XM_017004941.1 XM_017004942.1 XM_017004943.1 XM_017004944.1 XM_017004945.1 XM_017004946.1 XM_017004947.1 XM_017004948.1 XM_017004949.1 XM_017004950.1 XM_017004951.1 XM_017004952.1 XM_017004953.1 XM_017004954.1 XM_017004955.1 XM_017004956.1 XM_017004957.1 NR_110237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1007138 c.1262G>T p.R421I Chr.2: 143952013-143952013 on Build GRCh38
6466285 c.1202-1G>T p.? Chr.2: 143952074-143952074 on Build GRCh38
1399793 c.1240delA p.R414fs*12 Chr.2: 143952035-143952035 on Build GRCh38
6676653 c.1271T>G p.I424S Chr.2: 143952004-143952004 on Build GRCh38
7104345 c.1204G>A p.E402K Chr.2: 143952071-143952071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755345966 Chr.2: 143951944-143951944 on Build GRCh38
rs373283638 Chr.2: 143952091-143952091 on Build GRCh38
rs763920742 Chr.2: 143952004-143952004 on Build GRCh38
rs1038255359 Chr.2: 143951893-143951893 on Build GRCh38
rs748721916 Chr.2: 143951955-143951955 on Build GRCh38
rs780022414 Chr.2: 143952055-143952056 on Build GRCh38
rs371343368 Chr.2: 143952063-143952063 on Build GRCh38
rs749299854 Chr.2: 143951974-143951974 on Build GRCh38
rs555375276 Chr.2: 143952093-143952093 on Build GRCh38
rs902414666 Chr.2: 143951986-143951986 on Build GRCh38
rs142039455 Chr.2: 143952048-143952048 on Build GRCh38
rs906045977 Chr.2: 143951895-143951895 on Build GRCh38
rs546462806 Chr.2: 143951910-143951911 on Build GRCh38
rs770537850 Chr.2: 143951958-143951959 on Build GRCh38
rs940675066 Chr.2: 143951919-143951919 on Build GRCh38
rs1054906280 Chr.2: 143951911-143951911 on Build GRCh38
rs765231457 Chr.2: 143951910-143951910 on Build GRCh38
rs779751820 Chr.2: 143951941-143951941 on Build GRCh38
rs367849783 Chr.2: 143951993-143951993 on Build GRCh38
rs776237479 Chr.2: 143952041-143952041 on Build GRCh38
rs751191782 Chr.2: 143952092-143952092 on Build GRCh38
rs199546161 Chr.2: 143952107-143952107 on Build GRCh38
rs369423948 Chr.2: 143952086-143952086 on Build GRCh38
rs781588733 Chr.2: 143951945-143951945 on Build GRCh38
rs770239005 Chr.2: 143951965-143951965 on Build GRCh38
rs750017184 Chr.2: 143952073-143952073 on Build GRCh38
rs1030452432 Chr.2: 143952098-143952098 on Build GRCh38
rs754548644 Chr.2: 143952096-143952096 on Build GRCh38
rs773292975 Chr.2: 143951967-143951967 on Build GRCh38
rs747390311 Chr.2: 143951942-143951942 on Build GRCh38
rs758124368 Chr.2: 143951940-143951940 on Build GRCh38
rs1000802936 Chr.2: 143952039-143952039 on Build GRCh38
rs575778432 Chr.2: 143951995-143951995 on Build GRCh38
rs368490272 Chr.2: 143951978-143951978 on Build GRCh38

View Full Product Details