Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 5044444-5044671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf89
Entrez Gene ID
146556
Gene Name
chromosome 16 open reading frame 89
Gene Aliases
-
UniGene
Hs.11782
Gene Chromosome Location
Chr.16: 5042772-5066145 on Build GRCh38
Transcripts
NM_001098514.2 NM_152459.4 XM_005255143.3 XM_011522392.2 XM_017022972.1 XM_017022974.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667022 c.1312_1313insC p.V438fs*>4 Chr.16: 5044593-5044594 on Build GRCh38
6561845 c.1298C>T p.S433L Chr.16: 5044608-5044608 on Build GRCh38
1189176 c.1282A>G p.R428G Chr.16: 5044624-5044624 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563855081 Chr.16: 5044519-5044519 on Build GRCh38
rs769621981 Chr.16: 5044604-5044604 on Build GRCh38
rs759624493 Chr.16: 5044621-5044621 on Build GRCh38
rs773780962 Chr.16: 5044533-5044533 on Build GRCh38
rs561128734 Chr.16: 5044474-5044474 on Build GRCh38
rs777466700 Chr.16: 5044611-5044611 on Build GRCh38
rs947967064 Chr.16: 5044560-5044560 on Build GRCh38
rs8055684 Chr.16: 5044569-5044569 on Build GRCh38
rs532632376 Chr.16: 5044545-5044545 on Build GRCh38
rs966711510 Chr.16: 5044613-5044613 on Build GRCh38
rs753083090 Chr.16: 5044477-5044477 on Build GRCh38
rs28401183 Chr.16: 5044571-5044571 on Build GRCh38
rs751216403 Chr.16: 5044462-5044462 on Build GRCh38
rs979031459 Chr.16: 5044634-5044634 on Build GRCh38
rs764433165 Chr.16: 5044554-5044554 on Build GRCh38
rs780440686 Chr.16: 5044579-5044579 on Build GRCh38
rs767187950 Chr.16: 5044542-5044542 on Build GRCh38
rs756571043 Chr.16: 5044478-5044478 on Build GRCh38
rs754662565 Chr.16: 5044472-5044472 on Build GRCh38
rs747965111 Chr.16: 5044512-5044512 on Build GRCh38
rs367987886 Chr.16: 5044485-5044485 on Build GRCh38
rs746067105 Chr.16: 5044486-5044486 on Build GRCh38
rs775119793 Chr.16: 5044640-5044640 on Build GRCh38
rs778846405 Chr.16: 5044577-5044578 on Build GRCh38
rs776531500 Chr.16: 5044498-5044498 on Build GRCh38
rs534936133 Chr.16: 5044602-5044602 on Build GRCh38
rs758630776 Chr.16: 5044576-5044576 on Build GRCh38
rs967719357 Chr.16: 5044626-5044626 on Build GRCh38
rs372216879 Chr.16: 5044514-5044514 on Build GRCh38
rs770440494 Chr.16: 5044523-5044523 on Build GRCh38
rs548414551 Chr.16: 5044623-5044623 on Build GRCh38
rs773808493 Chr.16: 5044617-5044617 on Build GRCh38
rs761015465 Chr.16: 5044553-5044553 on Build GRCh38
rs774200208 Chr.16: 5044517-5044517 on Build GRCh38
rs772449266 Chr.16: 5044487-5044487 on Build GRCh38
rs1005464576 Chr.16: 5044578-5044578 on Build GRCh38
rs920331757 Chr.16: 5044644-5044644 on Build GRCh38
rs892425447 Chr.16: 5044483-5044483 on Build GRCh38
rs1049209416 Chr.16: 5044574-5044574 on Build GRCh38
rs940311454 Chr.16: 5044608-5044608 on Build GRCh38
rs567972068 Chr.16: 5044639-5044639 on Build GRCh38
rs183176637 Chr.16: 5044482-5044482 on Build GRCh38
rs375821911 Chr.16: 5044568-5044568 on Build GRCh38
rs747288641 Chr.16: 5044595-5044595 on Build GRCh38
rs749223336 Chr.16: 5044616-5044616 on Build GRCh38
rs370236561 Chr.16: 5044481-5044481 on Build GRCh38
rs759138882 Chr.16: 5044535-5044535 on Build GRCh38

View Full Product Details