Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64300930-64301155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK4-TEX40
Entrez Gene ID
106780802
Gene Name
KCNK4-TEX40 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 64291722-64304769 on Build GRCh38
Transcripts
NR_133662.1 NM_001039496.1

Gene Symbol
TEX40
Entrez Gene ID
25858
Gene Name
testis expressed 40
Gene Aliases
C11orf20
UniGene
Hs.97174
Gene Chromosome Location
Chr.11: 64300391-64304767 on Build GRCh38
Transcripts
NR_133662.1 NM_001039496.1

SNP Detail

SNP ID Chromosome Location
rs1008901952 Chr.11: 64301099-64301099 on Build GRCh38
rs955142961 Chr.11: 64301021-64301021 on Build GRCh38
rs559332912 Chr.11: 64301083-64301083 on Build GRCh38
rs375492502 Chr.11: 64300976-64300976 on Build GRCh38
rs549945778 Chr.11: 64301066-64301066 on Build GRCh38
rs570258786 Chr.11: 64301003-64301003 on Build GRCh38
rs372735141 Chr.11: 64300968-64300968 on Build GRCh38
rs758167931 Chr.11: 64300973-64300973 on Build GRCh38
rs964392174 Chr.11: 64301123-64301123 on Build GRCh38
rs776013464 Chr.11: 64301025-64301025 on Build GRCh38
rs912071059 Chr.11: 64301065-64301065 on Build GRCh38
rs975415490 Chr.11: 64301085-64301085 on Build GRCh38
rs768955786 Chr.11: 64301037-64301037 on Build GRCh38
rs946724199 Chr.11: 64301115-64301115 on Build GRCh38
rs563028561 Chr.11: 64301133-64301133 on Build GRCh38
rs986535550 Chr.11: 64301036-64301036 on Build GRCh38
rs142295121 Chr.11: 64301002-64301003 on Build GRCh38
rs539233589 Chr.11: 64301029-64301029 on Build GRCh38
rs753402158 Chr.11: 64300952-64300952 on Build GRCh38
rs566385281 Chr.11: 64301125-64301125 on Build GRCh38
rs112882340 Chr.11: 64300988-64300988 on Build GRCh38
rs781184785 Chr.11: 64300987-64300987 on Build GRCh38
rs891409833 Chr.11: 64301088-64301088 on Build GRCh38
rs186547485 Chr.11: 64300985-64300985 on Build GRCh38
rs898641374 Chr.11: 64300986-64300986 on Build GRCh38
rs915464620 Chr.11: 64301109-64301109 on Build GRCh38
rs779018105 Chr.11: 64300962-64300962 on Build GRCh38
rs770243190 Chr.11: 64301005-64301005 on Build GRCh38
rs774551524 Chr.11: 64300991-64300991 on Build GRCh38
rs1025312355 Chr.11: 64301020-64301020 on Build GRCh38

View Full Product Details