Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40659286-40659513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED14
Entrez Gene ID
9282
Gene Name
mediator complex subunit 14
Gene Aliases
CRSP150, CRSP2, CSRP, CXorf4, DRIP150, EXLM1, RGR1, TRAP170
UniGene
Hs.407604
Gene Chromosome Location
Chr.X: 40649543-40736122 on Build GRCh38
Transcripts
NM_004229.3 XM_005272701.2 XM_011544000.2 XM_017029962.1

SNP Detail

SNP ID Chromosome Location
rs772133009 Chr.X: 40659399-40659399 on Build GRCh38
rs377666861 Chr.X: 40659476-40659476 on Build GRCh38
rs749722802 Chr.X: 40659369-40659369 on Build GRCh38
rs374347911 Chr.X: 40659404-40659404 on Build GRCh38
rs771224723 Chr.X: 40659370-40659370 on Build GRCh38
rs757045701 Chr.X: 40659346-40659346 on Build GRCh38
rs759966437 Chr.X: 40659384-40659384 on Build GRCh38
rs986921035 Chr.X: 40659413-40659413 on Build GRCh38
rs774883112 Chr.X: 40659381-40659381 on Build GRCh38
rs147145468 Chr.X: 40659464-40659464 on Build GRCh38
rs778255544 Chr.X: 40659362-40659362 on Build GRCh38
rs371448258 Chr.X: 40659332-40659332 on Build GRCh38
rs773379004 Chr.X: 40659482-40659482 on Build GRCh38
rs750374710 Chr.X: 40659484-40659484 on Build GRCh38

View Full Product Details