Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57028702-57028975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO1A
Entrez Gene ID
4640
Gene Name
myosin IA
Gene Aliases
BBMI, DFNA48, MIHC, MYHL
UniGene
Hs.5394
Gene Chromosome Location
Chr.12: 57028517-57050765 on Build GRCh38
Transcripts
NM_001256041.1 NM_005379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6137400 c.3063A>T p.A1021A Chr.12: 57028824-57028824 on Build GRCh38
941854 c.3098A>G p.K1033R Chr.12: 57028789-57028789 on Build GRCh38
4765808 c.3099_3100insA p.G1034fs*>11 Chr.12: 57028787-57028788 on Build GRCh38
6720130 c.3086G>A p.R1029H Chr.12: 57028801-57028801 on Build GRCh38
6282952 c.3099G>T p.K1033N Chr.12: 57028788-57028788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567817417 Chr.12: 57028848-57028848 on Build GRCh38
rs766987099 Chr.12: 57028857-57028857 on Build GRCh38
rs759055851 Chr.12: 57028855-57028855 on Build GRCh38
rs762443700 Chr.12: 57028773-57028773 on Build GRCh38
rs769105633 Chr.12: 57028842-57028842 on Build GRCh38
rs922699155 Chr.12: 57028901-57028901 on Build GRCh38
rs751097838 Chr.12: 57028786-57028786 on Build GRCh38
rs1053291125 Chr.12: 57028956-57028956 on Build GRCh38
rs915521508 Chr.12: 57028892-57028892 on Build GRCh38
rs367774627 Chr.12: 57028801-57028801 on Build GRCh38
rs758444016 Chr.12: 57028837-57028837 on Build GRCh38
rs370617353 Chr.12: 57028759-57028759 on Build GRCh38
rs775638170 Chr.12: 57028747-57028747 on Build GRCh38
rs77915386 Chr.12: 57028792-57028792 on Build GRCh38
rs56347319 Chr.12: 57028944-57028944 on Build GRCh38
rs748756029 Chr.12: 57028788-57028789 on Build GRCh38
rs747253822 Chr.12: 57028839-57028839 on Build GRCh38
rs763817650 Chr.12: 57028907-57028907 on Build GRCh38
rs772324788 Chr.12: 57028740-57028740 on Build GRCh38
rs760324417 Chr.12: 57028896-57028896 on Build GRCh38
rs753706670 Chr.12: 57028808-57028808 on Build GRCh38
rs539337421 Chr.12: 57028948-57028948 on Build GRCh38
rs76394585 Chr.12: 57028861-57028861 on Build GRCh38
rs1014037608 Chr.12: 57028798-57028798 on Build GRCh38
rs745951847 Chr.12: 57028825-57028825 on Build GRCh38
rs535043825 Chr.12: 57028853-57028853 on Build GRCh38
rs750372080 Chr.12: 57028925-57028925 on Build GRCh38
rs922679479 Chr.12: 57028735-57028735 on Build GRCh38
rs547316446 Chr.12: 57028860-57028862 on Build GRCh38
rs368168684 Chr.12: 57028919-57028919 on Build GRCh38
rs956960371 Chr.12: 57028850-57028850 on Build GRCh38
rs753615797 Chr.12: 57028910-57028910 on Build GRCh38
rs763905867 Chr.12: 57028802-57028802 on Build GRCh38
rs944466855 Chr.12: 57028928-57028928 on Build GRCh38
rs201677711 Chr.12: 57028736-57028736 on Build GRCh38
rs371613423 Chr.12: 57028796-57028796 on Build GRCh38
rs766402013 Chr.12: 57028924-57028924 on Build GRCh38
rs373699223 Chr.12: 57028838-57028838 on Build GRCh38
rs748612876 Chr.12: 57028844-57028844 on Build GRCh38
rs765185808 Chr.12: 57028920-57028920 on Build GRCh38
rs777575982 Chr.12: 57028785-57028785 on Build GRCh38
rs770612489 Chr.12: 57028789-57028789 on Build GRCh38
rs910730261 Chr.12: 57028927-57028927 on Build GRCh38
rs370014993 Chr.12: 57028811-57028811 on Build GRCh38
rs148841648 Chr.12: 57028791-57028791 on Build GRCh38
rs961685802 Chr.12: 57028894-57028894 on Build GRCh38
rs367714595 Chr.12: 57028733-57028733 on Build GRCh38
rs145478643 Chr.12: 57028775-57028775 on Build GRCh38
rs976742743 Chr.12: 57028899-57028899 on Build GRCh38
rs760538634 Chr.12: 57028885-57028885 on Build GRCh38
rs974724392 Chr.12: 57028796-57028806 on Build GRCh38
rs374277812 Chr.12: 57028769-57028769 on Build GRCh38
rs986236886 Chr.12: 57028922-57028922 on Build GRCh38
rs777142608 Chr.12: 57028843-57028843 on Build GRCh38
rs778773547 Chr.12: 57028815-57028815 on Build GRCh38
rs564504774 Chr.12: 57028763-57028763 on Build GRCh38
rs936298255 Chr.12: 57028951-57028951 on Build GRCh38
rs758452894 Chr.12: 57028933-57028933 on Build GRCh38
rs944459094 Chr.12: 57028724-57028724 on Build GRCh38

View Full Product Details