Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157434691-157434922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTIP
Entrez Gene ID
9595
Gene Name
cytohesin 1 interacting protein
Gene Aliases
B3-1, CASP, CYBR, CYTHIP, HE, PSCDBP
UniGene
Hs.270
Gene Chromosome Location
Chr.2: 157412781-157444092 on Build GRCh38
Transcripts
NM_004288.4 XM_017005386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3895032 c.224G>A p.R75K Chr.2: 157434698-157434698 on Build GRCh38
7363591 c.220C>G p.Q74E Chr.2: 157434702-157434702 on Build GRCh38
6395355 c.216G>T p.W72C Chr.2: 157434706-157434706 on Build GRCh38
1249632 c.207C>T p.D69D Chr.2: 157434715-157434715 on Build GRCh38
5587592 c.197C>T p.S66F Chr.2: 157434725-157434725 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769634701 Chr.2: 157434817-157434822 on Build GRCh38
rs113065609 Chr.2: 157434813-157434820 on Build GRCh38
rs10536561 Chr.2: 157434849-157434849 on Build GRCh38
rs112950749 Chr.2: 157434853-157434853 on Build GRCh38
rs965904630 Chr.2: 157434837-157434837 on Build GRCh38
rs753980335 Chr.2: 157434732-157434732 on Build GRCh38
rs187839972 Chr.2: 157434777-157434777 on Build GRCh38
rs748830801 Chr.2: 157434792-157434792 on Build GRCh38
rs367942473 Chr.2: 157434845-157434845 on Build GRCh38
rs879166197 Chr.2: 157434813-157434814 on Build GRCh38
rs780886578 Chr.2: 157434762-157434762 on Build GRCh38
rs368427044 Chr.2: 157434813-157434818 on Build GRCh38
rs557688450 Chr.2: 157434725-157434725 on Build GRCh38
rs888877600 Chr.2: 157434836-157434861 on Build GRCh38
rs375314061 Chr.2: 157434843-157434843 on Build GRCh38
rs761916415 Chr.2: 157434744-157434744 on Build GRCh38
rs887443450 Chr.2: 157434820-157434820 on Build GRCh38
rs113623090 Chr.2: 157434851-157434851 on Build GRCh38
rs1049211611 Chr.2: 157434818-157434818 on Build GRCh38
rs752480411 Chr.2: 157434768-157434768 on Build GRCh38
rs112331549 Chr.2: 157434857-157434857 on Build GRCh38
rs751401151 Chr.2: 157434758-157434758 on Build GRCh38
rs112348634 Chr.2: 157434847-157434847 on Build GRCh38
rs368917412 Chr.2: 157434760-157434760 on Build GRCh38
rs751751424 Chr.2: 157434887-157434910 on Build GRCh38
rs1053387032 Chr.2: 157434890-157434890 on Build GRCh38
rs111713909 Chr.2: 157434855-157434855 on Build GRCh38
rs775893411 Chr.2: 157434718-157434718 on Build GRCh38
rs777344021 Chr.2: 157434782-157434782 on Build GRCh38
rs779647719 Chr.2: 157434800-157434800 on Build GRCh38
rs796639696 Chr.2: 157434844-157434857 on Build GRCh38
rs878928456 Chr.2: 157434842-157434861 on Build GRCh38
rs758077442 Chr.2: 157434749-157434749 on Build GRCh38
rs1007471688 Chr.2: 157434822-157434822 on Build GRCh38
rs997321682 Chr.2: 157434892-157434892 on Build GRCh38
rs368531225 Chr.2: 157434813-157434822 on Build GRCh38
rs897589915 Chr.2: 157434797-157434797 on Build GRCh38
rs764397531 Chr.2: 157434730-157434730 on Build GRCh38
rs1017458247 Chr.2: 157434824-157434824 on Build GRCh38
rs776347901 Chr.2: 157434815-157434822 on Build GRCh38
rs993224395 Chr.2: 157434812-157434812 on Build GRCh38
rs201946540 Chr.2: 157434750-157434750 on Build GRCh38
rs943097639 Chr.2: 157434838-157434838 on Build GRCh38
rs372228673 Chr.2: 157434813-157434816 on Build GRCh38
rs867002315 Chr.2: 157434859-157434859 on Build GRCh38

View Full Product Details