Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 120970436-120970669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLI2
Entrez Gene ID
2736
Gene Name
GLI family zinc finger 2
Gene Aliases
CJS, HPE9, PHS2, THP1, THP2
UniGene
Hs.111867
Gene Chromosome Location
Chr.2: 120797291-120992653 on Build GRCh38
Transcripts
NM_005270.4 XM_006712422.2 XM_011510969.2 XM_011510970.2 XM_011510971.2 XM_011510972.2 XM_011510973.2 XM_011510974.2 XM_017003818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3836701 c.964A>C p.T322P Chr.2: 120970511-120970511 on Build GRCh38
6671499 c.962C>T p.P321L Chr.2: 120970509-120970509 on Build GRCh38
1691000 c.1007C>A p.A336D Chr.2: 120970554-120970554 on Build GRCh38
7318554 c.909G>C p.Q303H Chr.2: 120970456-120970456 on Build GRCh38
4084549 c.981G>A p.Q327Q Chr.2: 120970528-120970528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745509639 Chr.2: 120970521-120970521 on Build GRCh38
rs776393864 Chr.2: 120970540-120970540 on Build GRCh38
rs767031465 Chr.2: 120970570-120970572 on Build GRCh38
rs755546739 Chr.2: 120970498-120970498 on Build GRCh38
rs761750550 Chr.2: 120970455-120970455 on Build GRCh38
rs763261944 Chr.2: 120970577-120970577 on Build GRCh38
rs997054628 Chr.2: 120970492-120970492 on Build GRCh38
rs768661123 Chr.2: 120970538-120970538 on Build GRCh38
rs149894186 Chr.2: 120970510-120970510 on Build GRCh38
rs141704487 Chr.2: 120970513-120970513 on Build GRCh38
rs764651661 Chr.2: 120970640-120970640 on Build GRCh38
rs372424926 Chr.2: 120970519-120970519 on Build GRCh38
rs765528898 Chr.2: 120970458-120970458 on Build GRCh38
rs748300953 Chr.2: 120970546-120970546 on Build GRCh38
rs773624892 Chr.2: 120970562-120970562 on Build GRCh38
rs201331588 Chr.2: 120970466-120970466 on Build GRCh38
rs773277858 Chr.2: 120970460-120970460 on Build GRCh38
rs377150486 Chr.2: 120970532-120970532 on Build GRCh38
rs749871248 Chr.2: 120970645-120970645 on Build GRCh38
rs367854976 Chr.2: 120970493-120970493 on Build GRCh38
rs768096911 Chr.2: 120970615-120970615 on Build GRCh38
rs369761915 Chr.2: 120970528-120970528 on Build GRCh38
rs766641468 Chr.2: 120970578-120970578 on Build GRCh38
rs753110716 Chr.2: 120970624-120970624 on Build GRCh38
rs775899835 Chr.2: 120970569-120970570 on Build GRCh38
rs756838422 Chr.2: 120970639-120970639 on Build GRCh38
rs144970612 Chr.2: 120970518-120970518 on Build GRCh38
rs1004316414 Chr.2: 120970515-120970515 on Build GRCh38
rs878981522 Chr.2: 120970630-120970639 on Build GRCh38
rs369580183 Chr.2: 120970610-120970610 on Build GRCh38
rs974103123 Chr.2: 120970605-120970605 on Build GRCh38
rs376694469 Chr.2: 120970646-120970646 on Build GRCh38
rs746753743 Chr.2: 120970536-120970536 on Build GRCh38
rs962710820 Chr.2: 120970558-120970558 on Build GRCh38
rs769741962 Chr.2: 120970547-120970547 on Build GRCh38
rs142356900 Chr.2: 120970550-120970550 on Build GRCh38
rs865966949 Chr.2: 120970488-120970488 on Build GRCh38
rs774869560 Chr.2: 120970603-120970603 on Build GRCh38
rs369035059 Chr.2: 120970647-120970647 on Build GRCh38
rs767850109 Chr.2: 120970509-120970509 on Build GRCh38
rs746686990 Chr.2: 120970487-120970487 on Build GRCh38

View Full Product Details