Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190659799-190660038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAB1
Entrez Gene ID
4664
Gene Name
NGFI-A binding protein 1
Gene Aliases
-
UniGene
Hs.107474
Gene Chromosome Location
Chr.2: 190648860-190692766 on Build GRCh38
Transcripts
NM_001321312.1 NM_001321313.1 NM_001321314.1 NM_001321315.1 NM_005966.3 XM_005246582.1 XM_005246583.1 XM_005246586.1 XM_005246587.1 XM_011511219.2 XM_017004170.1 XM_017004171.1 XM_017004172.1 XM_017004173.1 XM_017004174.1 XM_017004175.1 XM_017004176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6244109 c.709G>T p.E237* Chr.2: 190659885-190659885 on Build GRCh38
3575592 c.647A>G p.N216S Chr.2: 190659823-190659823 on Build GRCh38
1404118 c.766T>C p.Y256H Chr.2: 190659942-190659942 on Build GRCh38
2712628 c.644T>C p.L215S Chr.2: 190659820-190659820 on Build GRCh38
5973511 c.789G>A p.R263R Chr.2: 190659965-190659965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772600519 Chr.2: 190659835-190659835 on Build GRCh38
rs748510311 Chr.2: 190659830-190659830 on Build GRCh38
rs745371114 Chr.2: 190659852-190659852 on Build GRCh38
rs751648275 Chr.2: 190659906-190659906 on Build GRCh38
rs372366249 Chr.2: 190660007-190660007 on Build GRCh38
rs200545682 Chr.2: 190659836-190659836 on Build GRCh38
rs767473062 Chr.2: 190659902-190659902 on Build GRCh38
rs371357831 Chr.2: 190659990-190659990 on Build GRCh38
rs996090671 Chr.2: 190659874-190659874 on Build GRCh38
rs139792478 Chr.2: 190659986-190659986 on Build GRCh38
rs1053176 Chr.2: 190659924-190659924 on Build GRCh38
rs753091137 Chr.2: 190659981-190659981 on Build GRCh38
rs560393395 Chr.2: 190659967-190659967 on Build GRCh38
rs201301653 Chr.2: 190659892-190659892 on Build GRCh38
rs765427235 Chr.2: 190659976-190659976 on Build GRCh38
rs764864704 Chr.2: 190659959-190659960 on Build GRCh38
rs958139503 Chr.2: 190660005-190660005 on Build GRCh38
rs773236295 Chr.2: 190660012-190660012 on Build GRCh38
rs768291384 Chr.2: 190659881-190659881 on Build GRCh38
rs1053175 Chr.2: 190659907-190659907 on Build GRCh38
rs778211635 Chr.2: 190659846-190659846 on Build GRCh38
rs755220736 Chr.2: 190659960-190659960 on Build GRCh38
rs747526401 Chr.2: 190659987-190659987 on Build GRCh38
rs887340081 Chr.2: 190659886-190659886 on Build GRCh38
rs377284116 Chr.2: 190660006-190660006 on Build GRCh38
rs149297711 Chr.2: 190659929-190659929 on Build GRCh38
rs1024970469 Chr.2: 190659894-190659894 on Build GRCh38
rs748832731 Chr.2: 190659999-190659999 on Build GRCh38
rs775060146 Chr.2: 190659865-190659865 on Build GRCh38
rs766590461 Chr.2: 190659937-190659937 on Build GRCh38
rs771778876 Chr.2: 190660009-190660009 on Build GRCh38
rs777918335 Chr.2: 190659893-190659893 on Build GRCh38
rs769308730 Chr.2: 190659864-190659864 on Build GRCh38
rs779568155 Chr.2: 190659998-190659998 on Build GRCh38
rs1053174 Chr.2: 190659897-190659897 on Build GRCh38
rs375143739 Chr.2: 190660008-190660008 on Build GRCh38
rs762627880 Chr.2: 190659879-190659879 on Build GRCh38
rs754074907 Chr.2: 190659957-190659957 on Build GRCh38
rs368080938 Chr.2: 190659932-190659932 on Build GRCh38

View Full Product Details