Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 174448356-174448621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR52
Entrez Gene ID
9293
Gene Name
G protein-coupled receptor 52
Gene Aliases
-
UniGene
Hs.673850
Gene Chromosome Location
Chr.1: 174448074-174449545 on Build GRCh38
Transcripts
NM_005684.4 NM_014857.4 XM_005245680.1 XM_005245681.2 XM_006711693.1 XM_011510223.2 XM_017002994.1

Gene Symbol
RABGAP1L
Entrez Gene ID
9910
Gene Name
RAB GTPase activating protein 1 like
Gene Aliases
HHL, TBC1D18
UniGene
Hs.585378
Gene Chromosome Location
Chr.1: 174159414-174995308 on Build GRCh38
Transcripts
NM_005684.4 NM_014857.4 XM_005245680.1 XM_005245681.2 XM_006711693.1 XM_011510223.2 XM_017002994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7323879 c.367G>A p.V123I Chr.1: 174448478-174448478 on Build GRCh38
3863586 c.360C>T p.I120I Chr.1: 174448471-174448471 on Build GRCh38
1208720 c.368T>C p.V123A Chr.1: 174448479-174448479 on Build GRCh38
899867 c.476G>T p.R159I Chr.1: 174448587-174448587 on Build GRCh38
4025271 c.250G>A p.A84T Chr.1: 174448361-174448361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199740203 Chr.1: 174448478-174448478 on Build GRCh38
rs773876752 Chr.1: 174448514-174448514 on Build GRCh38
rs760528036 Chr.1: 174448523-174448523 on Build GRCh38
rs777938936 Chr.1: 174448588-174448588 on Build GRCh38
rs963368138 Chr.1: 174448506-174448506 on Build GRCh38
rs763702465 Chr.1: 174448395-174448395 on Build GRCh38
rs747544834 Chr.1: 174448454-174448454 on Build GRCh38
rs765304392 Chr.1: 174448559-174448559 on Build GRCh38
rs1038739312 Chr.1: 174448480-174448480 on Build GRCh38
rs147709761 Chr.1: 174448412-174448412 on Build GRCh38
rs997331133 Chr.1: 174448417-174448417 on Build GRCh38
rs748802654 Chr.1: 174448492-174448492 on Build GRCh38
rs140917627 Chr.1: 174448511-174448511 on Build GRCh38
rs142536057 Chr.1: 174448556-174448556 on Build GRCh38
rs999847740 Chr.1: 174448557-174448557 on Build GRCh38
rs769203005 Chr.1: 174448544-174448544 on Build GRCh38
rs76949857 Chr.1: 174448516-174448516 on Build GRCh38
rs778061724 Chr.1: 174448449-174448449 on Build GRCh38
rs758577089 Chr.1: 174448565-174448565 on Build GRCh38
rs950634775 Chr.1: 174448444-174448444 on Build GRCh38
rs774036652 Chr.1: 174448397-174448397 on Build GRCh38
rs761763151 Chr.1: 174448404-174448404 on Build GRCh38
rs916634525 Chr.1: 174448518-174448518 on Build GRCh38
rs368909262 Chr.1: 174448563-174448563 on Build GRCh38
rs1053161033 Chr.1: 174448580-174448580 on Build GRCh38
rs781708780 Chr.1: 174448481-174448481 on Build GRCh38
rs780857209 Chr.1: 174448415-174448417 on Build GRCh38
rs982327414 Chr.1: 174448527-174448527 on Build GRCh38
rs762493241 Chr.1: 174448379-174448379 on Build GRCh38
rs768205015 Chr.1: 174448499-174448499 on Build GRCh38
rs766391344 Chr.1: 174448425-174448425 on Build GRCh38
rs759541008 Chr.1: 174448542-174448542 on Build GRCh38
rs773184889 Chr.1: 174448520-174448520 on Build GRCh38
rs1016306663 Chr.1: 174448501-174448501 on Build GRCh38
rs141857283 Chr.1: 174448408-174448408 on Build GRCh38
rs779072161 Chr.1: 174448433-174448433 on Build GRCh38
rs533324072 Chr.1: 174448426-174448426 on Build GRCh38
rs758777410 Chr.1: 174448439-174448439 on Build GRCh38
rs150316785 Chr.1: 174448438-174448438 on Build GRCh38
rs747751170 Chr.1: 174448526-174448527 on Build GRCh38
rs141804847 Chr.1: 174448531-174448531 on Build GRCh38
rs755072219 Chr.1: 174448430-174448430 on Build GRCh38
rs761497182 Chr.1: 174448515-174448515 on Build GRCh38
rs267598179 Chr.1: 174448440-174448440 on Build GRCh38

View Full Product Details