Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89577376-89577623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_011523001.2 XM_017023138.1 XM_017023139.1 XM_017023140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4477291 c.213C>T p.P71P Chr.16: 89577577-89577577 on Build GRCh38
5423786 c.182G>C p.R61T Chr.16: 89577546-89577546 on Build GRCh38
1324222 c.198G>C p.R66R Chr.16: 89577562-89577562 on Build GRCh38
974902 c.255G>T p.E85D Chr.16: 89577619-89577619 on Build GRCh38
7216431 c.229T>C p.F77L Chr.16: 89577593-89577593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746899856 Chr.16: 89577577-89577577 on Build GRCh38
rs755955923 Chr.16: 89577585-89577585 on Build GRCh38
rs34344861 Chr.16: 89577550-89577550 on Build GRCh38
rs749587094 Chr.16: 89577597-89577597 on Build GRCh38
rs763486441 Chr.16: 89577529-89577529 on Build GRCh38
rs541488981 Chr.16: 89577462-89577462 on Build GRCh38
rs35701751 Chr.16: 89577589-89577589 on Build GRCh38
rs866038042 Chr.16: 89577485-89577485 on Build GRCh38
rs932603560 Chr.16: 89577518-89577518 on Build GRCh38
rs554788775 Chr.16: 89577446-89577446 on Build GRCh38
rs868758891 Chr.16: 89577470-89577470 on Build GRCh38
rs866303878 Chr.16: 89577551-89577551 on Build GRCh38
rs889570902 Chr.16: 89577521-89577521 on Build GRCh38
rs200538988 Chr.16: 89577510-89577510 on Build GRCh38
rs369107618 Chr.16: 89577561-89577561 on Build GRCh38
rs748171497 Chr.16: 89577583-89577583 on Build GRCh38
rs780555581 Chr.16: 89577491-89577491 on Build GRCh38
rs766823094 Chr.16: 89577492-89577492 on Build GRCh38
rs1035058086 Chr.16: 89577433-89577435 on Build GRCh38
rs774825737 Chr.16: 89577598-89577598 on Build GRCh38
rs946587922 Chr.16: 89577539-89577539 on Build GRCh38
rs1010948341 Chr.16: 89577569-89577569 on Build GRCh38
rs969459072 Chr.16: 89577447-89577447 on Build GRCh38
rs142649340 Chr.16: 89577463-89577463 on Build GRCh38
rs1023763085 Chr.16: 89577436-89577436 on Build GRCh38
rs577930324 Chr.16: 89577489-89577489 on Build GRCh38
rs369719899 Chr.16: 89577578-89577578 on Build GRCh38
rs561118662 Chr.16: 89577579-89577579 on Build GRCh38
rs771912937 Chr.16: 89577599-89577600 on Build GRCh38
rs188505272 Chr.16: 89577410-89577410 on Build GRCh38
rs199749974 Chr.16: 89577557-89577557 on Build GRCh38
rs879176029 Chr.16: 89577440-89577440 on Build GRCh38
rs752534377 Chr.16: 89577564-89577564 on Build GRCh38
rs1049657143 Chr.16: 89577520-89577520 on Build GRCh38
rs1031441411 Chr.16: 89577405-89577405 on Build GRCh38
rs960327189 Chr.16: 89577449-89577449 on Build GRCh38
rs536591292 Chr.16: 89577432-89577432 on Build GRCh38
rs455527 Chr.16: 89577593-89577593 on Build GRCh38
rs572985580 Chr.16: 89577452-89577452 on Build GRCh38
rs201422940 Chr.16: 89577499-89577499 on Build GRCh38
rs753293523 Chr.16: 89577536-89577536 on Build GRCh38
rs779183997 Chr.16: 89577505-89577505 on Build GRCh38
rs773840851 Chr.16: 89577453-89577453 on Build GRCh38
rs764815949 Chr.16: 89577560-89577560 on Build GRCh38
rs571255703 Chr.16: 89577602-89577602 on Build GRCh38
rs758126808 Chr.16: 89577565-89577565 on Build GRCh38
rs373791273 Chr.16: 89577572-89577572 on Build GRCh38

View Full Product Details