Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 142760194-142760447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIVEP2
Entrez Gene ID
3097
Gene Name
human immunodeficiency virus type I enhancer binding protein 2
Gene Aliases
HIV-EP2, MBP-2, MIBP1, MRD43, SHN2, ZAS2, ZNF40B
UniGene
Hs.510172
Gene Chromosome Location
Chr.6: 142751467-142945201 on Build GRCh38
Transcripts
NM_006734.3 XM_017010805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5447662 c.6053T>C p.I2018T Chr.6: 142760235-142760235 on Build GRCh38
4166911 c.6020C>T p.T2007M Chr.6: 142760268-142760268 on Build GRCh38
5585426 c.5872C>T p.P1958S Chr.6: 142760416-142760416 on Build GRCh38
5869446 c.5998A>G p.R2000G Chr.6: 142760290-142760290 on Build GRCh38
6680435 c.6019A>G p.T2007A Chr.6: 142760269-142760269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139323930 Chr.6: 142760267-142760267 on Build GRCh38
rs772803795 Chr.6: 142760422-142760422 on Build GRCh38
rs756540338 Chr.6: 142760304-142760304 on Build GRCh38
rs747938154 Chr.6: 142760410-142760410 on Build GRCh38
rs745823504 Chr.6: 142760253-142760253 on Build GRCh38
rs745886505 Chr.6: 142760321-142760321 on Build GRCh38
rs778100584 Chr.6: 142760309-142760309 on Build GRCh38
rs771864638 Chr.6: 142760257-142760257 on Build GRCh38
rs770118634 Chr.6: 142760223-142760223 on Build GRCh38
rs749509886 Chr.6: 142760226-142760226 on Build GRCh38
rs775610150 Chr.6: 142760350-142760350 on Build GRCh38
rs530358868 Chr.6: 142760312-142760312 on Build GRCh38
rs9373360 Chr.6: 142760236-142760236 on Build GRCh38
rs186682558 Chr.6: 142760320-142760320 on Build GRCh38
rs746928726 Chr.6: 142760352-142760352 on Build GRCh38
rs755951268 Chr.6: 142760253-142760258 on Build GRCh38
rs774240882 Chr.6: 142760242-142760242 on Build GRCh38
rs780223364 Chr.6: 142760402-142760402 on Build GRCh38
rs768674269 Chr.6: 142760409-142760409 on Build GRCh38
rs769597457 Chr.6: 142760419-142760419 on Build GRCh38
rs755526887 Chr.6: 142760281-142760281 on Build GRCh38
rs764450037 Chr.6: 142760390-142760390 on Build GRCh38
rs760597493 Chr.6: 142760264-142760264 on Build GRCh38
rs367677918 Chr.6: 142760423-142760423 on Build GRCh38
rs779055580 Chr.6: 142760394-142760394 on Build GRCh38
rs151011480 Chr.6: 142760392-142760392 on Build GRCh38
rs372188482 Chr.6: 142760429-142760429 on Build GRCh38
rs377164631 Chr.6: 142760268-142760268 on Build GRCh38
rs753757790 Chr.6: 142760285-142760285 on Build GRCh38
rs753168893 Chr.6: 142760303-142760303 on Build GRCh38
rs144077396 Chr.6: 142760368-142760368 on Build GRCh38
rs748460758 Chr.6: 142760219-142760219 on Build GRCh38
rs201231052 Chr.6: 142760293-142760293 on Build GRCh38
rs764743303 Chr.6: 142760366-142760366 on Build GRCh38
rs771104722 Chr.6: 142760239-142760239 on Build GRCh38
rs200019864 Chr.6: 142760396-142760396 on Build GRCh38
rs772171638 Chr.6: 142760332-142760332 on Build GRCh38
rs758464437 Chr.6: 142760400-142760400 on Build GRCh38
rs778012921 Chr.6: 142760224-142760224 on Build GRCh38
rs764807155 Chr.6: 142760278-142760278 on Build GRCh38
rs752547200 Chr.6: 142760407-142760407 on Build GRCh38
rs776500555 Chr.6: 142760358-142760358 on Build GRCh38
rs757522237 Chr.6: 142760393-142760393 on Build GRCh38
rs756560364 Chr.6: 142760388-142760388 on Build GRCh38
rs200310309 Chr.6: 142760387-142760387 on Build GRCh38
rs1047913555 Chr.6: 142760294-142760294 on Build GRCh38
rs768582619 Chr.6: 142760357-142760357 on Build GRCh38
rs772896640 Chr.6: 142760367-142760367 on Build GRCh38
rs775450593 Chr.6: 142760263-142760263 on Build GRCh38

View Full Product Details