Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 140238213-140238464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRP1B
Entrez Gene ID
53353
Gene Name
LDL receptor related protein 1B
Gene Aliases
LRP-1B, LRP-DIT, LRPDIT
UniGene
Hs.656461
Gene Chromosome Location
Chr.2: 140231423-142132463 on Build GRCh38
Transcripts
NM_018557.2 XM_017004341.1 XM_017004342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5491486 c.13464A>G p.V4488V Chr.2: 140238248-140238248 on Build GRCh38
329493 c.13425A>C p.T4475T Chr.2: 140238287-140238287 on Build GRCh38
1006980 c.13443T>G p.I4481M Chr.2: 140238269-140238269 on Build GRCh38
1305668 c.13495G>A p.E4499K Chr.2: 140238217-140238217 on Build GRCh38
3567101 c.13450G>A p.G4484R Chr.2: 140238262-140238262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046389211 Chr.2: 140238382-140238382 on Build GRCh38
rs1013750952 Chr.2: 140238318-140238318 on Build GRCh38
rs145781835 Chr.2: 140238286-140238286 on Build GRCh38
rs764487280 Chr.2: 140238314-140238314 on Build GRCh38
rs75758215 Chr.2: 140238274-140238274 on Build GRCh38
rs773803360 Chr.2: 140238269-140238269 on Build GRCh38
rs963580455 Chr.2: 140238402-140238402 on Build GRCh38
rs745380235 Chr.2: 140238331-140238331 on Build GRCh38
rs568825566 Chr.2: 140238357-140238357 on Build GRCh38
rs771740614 Chr.2: 140238343-140238346 on Build GRCh38
rs775235846 Chr.2: 140238299-140238299 on Build GRCh38
rs908559066 Chr.2: 140238329-140238329 on Build GRCh38
rs761154055 Chr.2: 140238310-140238310 on Build GRCh38
rs754410321 Chr.2: 140238316-140238316 on Build GRCh38
rs1016331649 Chr.2: 140238398-140238398 on Build GRCh38
rs200181272 Chr.2: 140238249-140238249 on Build GRCh38
rs974565292 Chr.2: 140238425-140238426 on Build GRCh38
rs983968192 Chr.2: 140238397-140238397 on Build GRCh38
rs921727888 Chr.2: 140238437-140238437 on Build GRCh38
rs201081715 Chr.2: 140238341-140238341 on Build GRCh38

View Full Product Details