Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41397530-41397773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MX2
Entrez Gene ID
4600
Gene Name
MX dynamin like GTPase 2
Gene Aliases
MXB
UniGene
Hs.926
Gene Chromosome Location
Chr.21: 41361986-41408943 on Build GRCh38
Transcripts
NM_002463.1 XM_005260983.4 XM_005260984.1 XM_011529571.1 XM_011529572.1 XM_011529573.1 XM_011529574.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6094617 c.1140G>T p.M380I Chr.21: 41397682-41397682 on Build GRCh38
4634606 c.1086G>T p.E362D Chr.21: 41397628-41397628 on Build GRCh38
3550934 c.1100T>C p.V367A Chr.21: 41397642-41397642 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761404065 Chr.21: 41397585-41397585 on Build GRCh38
rs890001630 Chr.21: 41397591-41397591 on Build GRCh38
rs752127303 Chr.21: 41397612-41397612 on Build GRCh38
rs772004219 Chr.21: 41397649-41397649 on Build GRCh38
rs760005073 Chr.21: 41397700-41397700 on Build GRCh38
rs755024021 Chr.21: 41397602-41397602 on Build GRCh38
rs139898288 Chr.21: 41397671-41397671 on Build GRCh38
rs938765580 Chr.21: 41397677-41397677 on Build GRCh38
rs745812664 Chr.21: 41397647-41397647 on Build GRCh38
rs746796616 Chr.21: 41397666-41397666 on Build GRCh38
rs754069389 Chr.21: 41397730-41397730 on Build GRCh38
rs753065805 Chr.21: 41397710-41397710 on Build GRCh38
rs199541902 Chr.21: 41397668-41397668 on Build GRCh38
rs370561843 Chr.21: 41397605-41397605 on Build GRCh38
rs149795199 Chr.21: 41397692-41397692 on Build GRCh38
rs1030925519 Chr.21: 41397610-41397610 on Build GRCh38
rs753076753 Chr.21: 41397633-41397633 on Build GRCh38
rs760274916 Chr.21: 41397565-41397565 on Build GRCh38
rs374003780 Chr.21: 41397639-41397639 on Build GRCh38
rs764372288 Chr.21: 41397721-41397721 on Build GRCh38
rs754486957 Chr.21: 41397562-41397563 on Build GRCh38
rs757329930 Chr.21: 41397641-41397641 on Build GRCh38
rs897540294 Chr.21: 41397751-41397751 on Build GRCh38
rs775304711 Chr.21: 41397563-41397563 on Build GRCh38
rs981644297 Chr.21: 41397560-41397561 on Build GRCh38
rs1014546712 Chr.21: 41397749-41397749 on Build GRCh38
rs748381668 Chr.21: 41397617-41397617 on Build GRCh38
rs775187818 Chr.21: 41397653-41397653 on Build GRCh38
rs375985045 Chr.21: 41397566-41397566 on Build GRCh38
rs756500413 Chr.21: 41397713-41397713 on Build GRCh38
rs1033149222 Chr.21: 41397625-41397625 on Build GRCh38
rs1030333249 Chr.21: 41397570-41397570 on Build GRCh38
rs150660002 Chr.21: 41397616-41397616 on Build GRCh38
rs896329946 Chr.21: 41397693-41397695 on Build GRCh38
rs989007244 Chr.21: 41397676-41397676 on Build GRCh38
rs1005874188 Chr.21: 41397562-41397562 on Build GRCh38
rs183275800 Chr.21: 41397553-41397553 on Build GRCh38
rs370154196 Chr.21: 41397701-41397701 on Build GRCh38
rs776329916 Chr.21: 41397580-41397580 on Build GRCh38
rs962142972 Chr.21: 41397597-41397597 on Build GRCh38
rs953625578 Chr.21: 41397613-41397613 on Build GRCh38
rs1056264694 Chr.21: 41397689-41397689 on Build GRCh38
rs778904108 Chr.21: 41397644-41397644 on Build GRCh38
rs770694990 Chr.21: 41397640-41397640 on Build GRCh38
rs367788793 Chr.21: 41397648-41397648 on Build GRCh38
rs761499521 Chr.21: 41397678-41397678 on Build GRCh38
rs772810358 Chr.21: 41397693-41397693 on Build GRCh38

View Full Product Details