Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 11608232-11608461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
UniGene
Hs.567259
Gene Chromosome Location
Chr.17: 11598431-11970168 on Build GRCh38
Transcripts
NM_001372.3 XM_011523703.1 XM_017024292.1 XM_017024294.1 XM_017024295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4972397 c.607G>A p.E203K Chr.17: 11608318-11608318 on Build GRCh38
3889275 c.545G>A p.G182E Chr.17: 11608256-11608256 on Build GRCh38
6556181 c.532G>T p.E178* Chr.17: 11608243-11608243 on Build GRCh38
5524855 c.577G>A p.E193K Chr.17: 11608288-11608288 on Build GRCh38
6532394 c.530T>C p.L177P Chr.17: 11608241-11608241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992090840 Chr.17: 11608431-11608431 on Build GRCh38
rs775580134 Chr.17: 11608354-11608354 on Build GRCh38
rs749717578 Chr.17: 11608334-11608334 on Build GRCh38
rs776278316 Chr.17: 11608373-11608373 on Build GRCh38
rs758030440 Chr.17: 11608299-11608299 on Build GRCh38
rs750072255 Chr.17: 11608313-11608313 on Build GRCh38
rs764315084 Chr.17: 11608368-11608368 on Build GRCh38
rs1032111093 Chr.17: 11608420-11608420 on Build GRCh38
rs1010650271 Chr.17: 11608345-11608345 on Build GRCh38
rs754168239 Chr.17: 11608288-11608289 on Build GRCh38
rs200109759 Chr.17: 11608259-11608259 on Build GRCh38
rs771333243 Chr.17: 11608335-11608335 on Build GRCh38
rs749897988 Chr.17: 11608296-11608296 on Build GRCh38
rs765277789 Chr.17: 11608294-11608294 on Build GRCh38
rs1005957435 Chr.17: 11608364-11608364 on Build GRCh38
rs953799453 Chr.17: 11608433-11608433 on Build GRCh38
rs760821470 Chr.17: 11608362-11608362 on Build GRCh38
rs144614487 Chr.17: 11608432-11608432 on Build GRCh38
rs148474824 Chr.17: 11608440-11608440 on Build GRCh38
rs962008135 Chr.17: 11608426-11608428 on Build GRCh38
rs757086800 Chr.17: 11608268-11608268 on Build GRCh38
rs573879544 Chr.17: 11608415-11608415 on Build GRCh38
rs969261781 Chr.17: 11608434-11608434 on Build GRCh38
rs755883355 Chr.17: 11608353-11608353 on Build GRCh38
rs761735351 Chr.17: 11608374-11608374 on Build GRCh38
rs779732832 Chr.17: 11608304-11608304 on Build GRCh38
rs1000666870 Chr.17: 11608359-11608359 on Build GRCh38
rs371734467 Chr.17: 11608350-11608350 on Build GRCh38
rs555588377 Chr.17: 11608324-11608324 on Build GRCh38
rs374405924 Chr.17: 11608305-11608305 on Build GRCh38
rs753673564 Chr.17: 11608266-11608266 on Build GRCh38
rs754680221 Chr.17: 11608312-11608312 on Build GRCh38
rs745916988 Chr.17: 11608351-11608351 on Build GRCh38
rs757635352 Chr.17: 11608301-11608302 on Build GRCh38
rs985141282 Chr.17: 11608437-11608437 on Build GRCh38

View Full Product Details