Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 122862357-122862618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAM
Entrez Gene ID
56253
Gene Name
cytotoxic and regulatory T-cell molecule
Gene Aliases
CD355
UniGene
Hs.159523
Gene Chromosome Location
Chr.11: 122838419-122872643 on Build GRCh38
Transcripts
NM_001304782.1 NM_019604.3 XM_011542900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522440 c.689G>A p.R230K Chr.11: 122862500-122862500 on Build GRCh38
5961781 c.712C>G p.P238A Chr.11: 122862523-122862523 on Build GRCh38
5961782 c.115C>G p.P39A Chr.11: 122862523-122862523 on Build GRCh38
5776954 c.702C>T p.S234S Chr.11: 122862513-122862513 on Build GRCh38
3359057 c.94A>G p.N32D Chr.11: 122862502-122862502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763317059 Chr.11: 122862536-122862536 on Build GRCh38
rs765707849 Chr.11: 122862523-122862523 on Build GRCh38
rs766794474 Chr.11: 122862537-122862537 on Build GRCh38
rs755203596 Chr.11: 122862550-122862550 on Build GRCh38
rs781378057 Chr.11: 122862552-122862552 on Build GRCh38
rs756165854 Chr.11: 122862557-122862557 on Build GRCh38
rs912225103 Chr.11: 122862379-122862379 on Build GRCh38
rs370998914 Chr.11: 122862462-122862462 on Build GRCh38
rs144434993 Chr.11: 122862513-122862513 on Build GRCh38
rs201112405 Chr.11: 122862586-122862586 on Build GRCh38
rs755150231 Chr.11: 122862422-122862422 on Build GRCh38
rs771839876 Chr.11: 122862574-122862574 on Build GRCh38
rs868634130 Chr.11: 122862463-122862463 on Build GRCh38
rs202149204 Chr.11: 122862582-122862582 on Build GRCh38
rs367559366 Chr.11: 122862585-122862585 on Build GRCh38
rs373800697 Chr.11: 122862553-122862553 on Build GRCh38
rs776342253 Chr.11: 122862507-122862507 on Build GRCh38
rs751854056 Chr.11: 122862546-122862546 on Build GRCh38
rs750915787 Chr.11: 122862527-122862527 on Build GRCh38
rs898555053 Chr.11: 122862387-122862387 on Build GRCh38
rs758755606 Chr.11: 122862399-122862399 on Build GRCh38
rs747275256 Chr.11: 122862409-122862409 on Build GRCh38
rs781234454 Chr.11: 122862447-122862447 on Build GRCh38
rs745851877 Chr.11: 122862573-122862573 on Build GRCh38
rs748166383 Chr.11: 122862431-122862431 on Build GRCh38
rs569819710 Chr.11: 122862496-122862496 on Build GRCh38
rs149701020 Chr.11: 122862487-122862487 on Build GRCh38
rs943692053 Chr.11: 122862386-122862386 on Build GRCh38
rs777818588 Chr.11: 122862554-122862554 on Build GRCh38
rs780184666 Chr.11: 122862407-122862407 on Build GRCh38
rs754910695 Chr.11: 122862435-122862435 on Build GRCh38
rs774339987 Chr.11: 122862453-122862453 on Build GRCh38
rs781252243 Chr.11: 122862430-122862430 on Build GRCh38
rs770913459 Chr.11: 122862437-122862437 on Build GRCh38
rs374861787 Chr.11: 122862455-122862455 on Build GRCh38
rs777665107 Chr.11: 122862558-122862558 on Build GRCh38
rs139514014 Chr.11: 122862479-122862479 on Build GRCh38
rs750823922 Chr.11: 122862392-122862392 on Build GRCh38

View Full Product Details