Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155285607-155285838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN3
Entrez Gene ID
57657
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Gene Aliases
-
UniGene
Hs.706960
Gene Chromosome Location
Chr.1: 155277427-155289848 on Build GRCh38
Transcripts
NM_020897.2 NR_073074.1 XM_011509816.2 XM_011509817.2 XM_011509818.2 XM_017001918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668030 c.1251C>A p.F417L Chr.1: 155285738-155285738 on Build GRCh38
7212908 c.1249T>C p.F417L Chr.1: 155285736-155285736 on Build GRCh38
4936450 c.1351C>A p.P451T Chr.1: 155285838-155285838 on Build GRCh38
363034 c.1333C>T p.R445C Chr.1: 155285820-155285820 on Build GRCh38
1334611 c.1288G>A p.A430T Chr.1: 155285775-155285775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781614213 Chr.1: 155285806-155285806 on Build GRCh38
rs1009948050 Chr.1: 155285758-155285758 on Build GRCh38
rs561287940 Chr.1: 155285732-155285732 on Build GRCh38
rs996180637 Chr.1: 155285629-155285629 on Build GRCh38
rs893819521 Chr.1: 155285746-155285746 on Build GRCh38
rs1041276426 Chr.1: 155285716-155285716 on Build GRCh38
rs547981159 Chr.1: 155285655-155285655 on Build GRCh38
rs552224588 Chr.1: 155285707-155285707 on Build GRCh38
rs143562699 Chr.1: 155285763-155285763 on Build GRCh38
rs754707909 Chr.1: 155285717-155285717 on Build GRCh38
rs770400191 Chr.1: 155285815-155285815 on Build GRCh38
rs1033228902 Chr.1: 155285675-155285675 on Build GRCh38
rs773894483 Chr.1: 155285816-155285816 on Build GRCh38
rs991126266 Chr.1: 155285688-155285688 on Build GRCh38
rs759132143 Chr.1: 155285742-155285742 on Build GRCh38
rs34218829 Chr.1: 155285819-155285819 on Build GRCh38
rs748516371 Chr.1: 155285808-155285808 on Build GRCh38
rs769028932 Chr.1: 155285702-155285702 on Build GRCh38
rs919625510 Chr.1: 155285636-155285636 on Build GRCh38
rs747193196 Chr.1: 155285783-155285783 on Build GRCh38
rs1028970500 Chr.1: 155285642-155285642 on Build GRCh38
rs372807362 Chr.1: 155285779-155285779 on Build GRCh38
rs1020354904 Chr.1: 155285662-155285662 on Build GRCh38
rs752440221 Chr.1: 155285748-155285748 on Build GRCh38
rs747348182 Chr.1: 155285679-155285679 on Build GRCh38
rs797022182 Chr.1: 155285768-155285768 on Build GRCh38
rs780987766 Chr.1: 155285740-155285740 on Build GRCh38
rs552426994 Chr.1: 155285665-155285665 on Build GRCh38
rs944190536 Chr.1: 155285695-155285695 on Build GRCh38
rs751217415 Chr.1: 155285735-155285735 on Build GRCh38
rs762311126 Chr.1: 155285710-155285710 on Build GRCh38
rs770268384 Chr.1: 155285728-155285728 on Build GRCh38
rs978952533 Chr.1: 155285666-155285666 on Build GRCh38
rs772832771 Chr.1: 155285730-155285730 on Build GRCh38
rs146290600 Chr.1: 155285820-155285820 on Build GRCh38
rs899995641 Chr.1: 155285718-155285718 on Build GRCh38
rs750482495 Chr.1: 155285767-155285767 on Build GRCh38
rs755154997 Chr.1: 155285796-155285796 on Build GRCh38
rs118001822 Chr.1: 155285656-155285656 on Build GRCh38
rs576680898 Chr.1: 155285745-155285745 on Build GRCh38

View Full Product Details