Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 40688345-40688618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAFB
Entrez Gene ID
9935
Gene Name
MAF bZIP transcription factor B
Gene Aliases
KRML, MCTO
UniGene
Hs.169487
Gene Chromosome Location
Chr.20: 40685848-40689240 on Build GRCh38
Transcripts
NM_005461.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4098367 c.416C>A p.P139H Chr.20: 40688435-40688435 on Build GRCh38
5817122 c.362T>C p.L121P Chr.20: 40688489-40688489 on Build GRCh38
4098369 c.309G>A p.T103T Chr.20: 40688542-40688542 on Build GRCh38
478150 c.437C>T p.P146L Chr.20: 40688414-40688414 on Build GRCh38
4098368 c.387C>T p.G129G Chr.20: 40688464-40688464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552064209 Chr.20: 40688563-40688563 on Build GRCh38
rs781078964 Chr.20: 40688388-40688388 on Build GRCh38
rs201783302 Chr.20: 40688562-40688562 on Build GRCh38
rs141490664 Chr.20: 40688495-40688496 on Build GRCh38
rs371814365 Chr.20: 40688446-40688446 on Build GRCh38
rs551111140 Chr.20: 40688407-40688407 on Build GRCh38
rs952100058 Chr.20: 40688447-40688455 on Build GRCh38
rs879255276 Chr.20: 40688411-40688411 on Build GRCh38
rs1025517387 Chr.20: 40688554-40688554 on Build GRCh38
rs746418143 Chr.20: 40688509-40688509 on Build GRCh38
rs375150121 Chr.20: 40688462-40688462 on Build GRCh38
rs757701275 Chr.20: 40688502-40688502 on Build GRCh38
rs749649554 Chr.20: 40688412-40688412 on Build GRCh38
rs771626150 Chr.20: 40688419-40688419 on Build GRCh38
rs571395644 Chr.20: 40688422-40688422 on Build GRCh38
rs762729675 Chr.20: 40688452-40688452 on Build GRCh38
rs780801645 Chr.20: 40688510-40688510 on Build GRCh38
rs545827728 Chr.20: 40688497-40688497 on Build GRCh38
rs374174345 Chr.20: 40688557-40688557 on Build GRCh38
rs756368724 Chr.20: 40688403-40688403 on Build GRCh38
rs893483748 Chr.20: 40688394-40688394 on Build GRCh38
rs769412741 Chr.20: 40688449-40688449 on Build GRCh38
rs769500671 Chr.20: 40688514-40688514 on Build GRCh38
rs775390598 Chr.20: 40688429-40688430 on Build GRCh38
rs756456686 Chr.20: 40688491-40688491 on Build GRCh38
rs150718854 Chr.20: 40688511-40688511 on Build GRCh38
rs752988943 Chr.20: 40688392-40688392 on Build GRCh38
rs121912307 Chr.20: 40688458-40688458 on Build GRCh38
rs764399516 Chr.20: 40688492-40688492 on Build GRCh38
rs768735706 Chr.20: 40688446-40688447 on Build GRCh38
rs775963285 Chr.20: 40688443-40688443 on Build GRCh38
rs752691658 Chr.20: 40688489-40688489 on Build GRCh38
rs759347042 Chr.20: 40688475-40688475 on Build GRCh38
rs367955583 Chr.20: 40688428-40688428 on Build GRCh38
rs773047325 Chr.20: 40688450-40688450 on Build GRCh38
rs779090359 Chr.20: 40688506-40688506 on Build GRCh38
rs767671011 Chr.20: 40688483-40688483 on Build GRCh38
rs755086273 Chr.20: 40688371-40688371 on Build GRCh38
rs1045021347 Chr.20: 40688515-40688515 on Build GRCh38
rs762632933 Chr.20: 40688447-40688449 on Build GRCh38

View Full Product Details