Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 28934990-28935257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTN1
Entrez Gene ID
26046
Gene Name
listerin E3 ubiquitin protein ligase 1
Gene Aliases
C21orf10, C21orf98, RNF160, ZNF294
UniGene
Hs.288773
Gene Chromosome Location
Chr.21: 28928144-28992959 on Build GRCh38
Transcripts
NM_001320766.1 NM_015565.2 XM_017028316.1 XM_017028317.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5597038 c.4825C>T p.Q1609* Chr.21: 28935159-28935159 on Build GRCh38
5015689 c.4799A>T p.K1600M Chr.21: 28935185-28935185 on Build GRCh38
1223922 c.4757G>A p.S1586N Chr.21: 28935227-28935227 on Build GRCh38
3740261 c.4791T>C p.F1597F Chr.21: 28935193-28935193 on Build GRCh38
7291603 c.4765C>T p.R1589C Chr.21: 28935219-28935219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372185078 Chr.21: 28935149-28935149 on Build GRCh38
rs773770565 Chr.21: 28935150-28935150 on Build GRCh38
rs767130365 Chr.21: 28935173-28935173 on Build GRCh38
rs536009144 Chr.21: 28935046-28935046 on Build GRCh38
rs757622740 Chr.21: 28935187-28935187 on Build GRCh38
rs749270715 Chr.21: 28935200-28935200 on Build GRCh38
rs994790082 Chr.21: 28935056-28935056 on Build GRCh38
rs765675804 Chr.21: 28935201-28935201 on Build GRCh38
rs780090699 Chr.21: 28935077-28935077 on Build GRCh38
rs373620814 Chr.21: 28935060-28935060 on Build GRCh38
rs750951641 Chr.21: 28935205-28935205 on Build GRCh38
rs971512669 Chr.21: 28935218-28935218 on Build GRCh38
rs112621155 Chr.21: 28935175-28935175 on Build GRCh38
rs866586913 Chr.21: 28935124-28935124 on Build GRCh38
rs754225381 Chr.21: 28935184-28935184 on Build GRCh38
rs143896676 Chr.21: 28935133-28935133 on Build GRCh38
rs779463880 Chr.21: 28935067-28935067 on Build GRCh38
rs769853275 Chr.21: 28935127-28935127 on Build GRCh38
rs367755686 Chr.21: 28935110-28935110 on Build GRCh38
rs745881497 Chr.21: 28935069-28935069 on Build GRCh38
rs147275574 Chr.21: 28935142-28935142 on Build GRCh38
rs1045571367 Chr.21: 28935206-28935206 on Build GRCh38
rs758434881 Chr.21: 28935231-28935231 on Build GRCh38
rs759203373 Chr.21: 28935168-28935168 on Build GRCh38
rs879060842 Chr.21: 28935048-28935048 on Build GRCh38
rs375380804 Chr.21: 28935075-28935075 on Build GRCh38
rs747289384 Chr.21: 28935082-28935082 on Build GRCh38
rs780153221 Chr.21: 28935232-28935232 on Build GRCh38
rs768802582 Chr.21: 28935096-28935096 on Build GRCh38
rs576818321 Chr.21: 28935026-28935026 on Build GRCh38
rs371452182 Chr.21: 28935062-28935062 on Build GRCh38
rs776468458 Chr.21: 28935102-28935102 on Build GRCh38
rs773291855 Chr.21: 28935130-28935130 on Build GRCh38
rs917075560 Chr.21: 28935179-28935179 on Build GRCh38

View Full Product Details