Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60700740-60701001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A8
Entrez Gene ID
83661
Gene Name
membrane spanning 4-domains A8
Gene Aliases
4SPAN4, CD20L5, MS4A4, MS4A8B
UniGene
Hs.150878
Gene Chromosome Location
Chr.11: 60699574-60715811 on Build GRCh38
Transcripts
NM_031457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4560342 c.83G>A p.G28E Chr.11: 60700943-60700943 on Build GRCh38
7232622 c.26C>G p.P9R Chr.11: 60700886-60700886 on Build GRCh38
124510 c.64G>T p.G22C Chr.11: 60700924-60700924 on Build GRCh38
3953528 c.94C>G p.H32D Chr.11: 60700954-60700954 on Build GRCh38
5698193 c.30G>T p.V10V Chr.11: 60700890-60700890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754630693 Chr.11: 60700820-60700820 on Build GRCh38
rs200254161 Chr.11: 60700938-60700938 on Build GRCh38
rs762932701 Chr.11: 60700906-60700906 on Build GRCh38
rs751351627 Chr.11: 60700813-60700813 on Build GRCh38
rs892969862 Chr.11: 60700944-60700944 on Build GRCh38
rs200962516 Chr.11: 60700869-60700869 on Build GRCh38
rs183590907 Chr.11: 60700957-60700957 on Build GRCh38
rs149106877 Chr.11: 60700962-60700962 on Build GRCh38
rs768808826 Chr.11: 60700881-60700881 on Build GRCh38
rs934774593 Chr.11: 60700792-60700792 on Build GRCh38
rs758137123 Chr.11: 60700901-60700903 on Build GRCh38
rs766101464 Chr.11: 60700810-60700810 on Build GRCh38
rs201880847 Chr.11: 60700842-60700842 on Build GRCh38
rs372626395 Chr.11: 60700851-60700851 on Build GRCh38
rs746063389 Chr.11: 60700778-60700778 on Build GRCh38
rs947417644 Chr.11: 60700821-60700821 on Build GRCh38
rs776863093 Chr.11: 60700886-60700886 on Build GRCh38
rs758758486 Chr.11: 60700922-60700922 on Build GRCh38
rs778208143 Chr.11: 60700930-60700930 on Build GRCh38
rs952250897 Chr.11: 60700795-60700795 on Build GRCh38
rs764484076 Chr.11: 60700939-60700939 on Build GRCh38
rs746321484 Chr.11: 60700846-60700846 on Build GRCh38
rs145052992 Chr.11: 60700960-60700960 on Build GRCh38
rs753395781 Chr.11: 60700973-60700973 on Build GRCh38
rs748256385 Chr.11: 60700887-60700887 on Build GRCh38
rs767109802 Chr.11: 60700952-60700952 on Build GRCh38
rs143047257 Chr.11: 60700982-60700982 on Build GRCh38
rs770893882 Chr.11: 60700915-60700915 on Build GRCh38
rs751725725 Chr.11: 60700956-60700956 on Build GRCh38
rs767180589 Chr.11: 60700823-60700823 on Build GRCh38
rs141281628 Chr.11: 60700878-60700878 on Build GRCh38
rs765309013 Chr.11: 60700831-60700831 on Build GRCh38
rs1053123333 Chr.11: 60700793-60700793 on Build GRCh38
rs202135543 Chr.11: 60700976-60700976 on Build GRCh38
rs763470038 Chr.11: 60700790-60700790 on Build GRCh38
rs1039665553 Chr.11: 60700830-60700830 on Build GRCh38
rs773143727 Chr.11: 60700890-60700890 on Build GRCh38
rs374580601 Chr.11: 60700866-60700866 on Build GRCh38
rs752664736 Chr.11: 60700885-60700885 on Build GRCh38
rs752174575 Chr.11: 60700840-60700840 on Build GRCh38
rs769797287 Chr.11: 60700889-60700889 on Build GRCh38

View Full Product Details