Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35738239-35738480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA2
Entrez Gene ID
57704
Gene Name
glucosylceramidase beta 2
Gene Aliases
AD035, NLGase, SPG46
UniGene
Hs.443134
Gene Chromosome Location
Chr.9: 35736862-35749255 on Build GRCh38
Transcripts
NM_020944.2 XM_005251526.4 XM_006716809.3 XM_011517973.2 XM_017014937.1 XM_017014938.1 XM_017014939.1 XM_017014940.1 XM_017014941.1 XM_017014942.1 XM_017014943.1 XM_017014944.1 XM_017014945.1 XM_017014946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7096137 c.2157C>T p.L719L Chr.9: 35738272-35738272 on Build GRCh38
1674484 c.2120G>T p.G707V Chr.9: 35738309-35738309 on Build GRCh38
4870549 c.2089C>A p.L697M Chr.9: 35738358-35738358 on Build GRCh38
4889775 c.2150C>G p.S717C Chr.9: 35738279-35738279 on Build GRCh38
3745994 c.2145T>G p.F715L Chr.9: 35738284-35738284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960041201 Chr.9: 35738296-35738296 on Build GRCh38
rs113721604 Chr.9: 35738263-35738263 on Build GRCh38
rs911416899 Chr.9: 35738454-35738454 on Build GRCh38
rs757918567 Chr.9: 35738270-35738270 on Build GRCh38
rs774656141 Chr.9: 35738363-35738363 on Build GRCh38
rs761919429 Chr.9: 35738444-35738444 on Build GRCh38
rs146034295 Chr.9: 35738271-35738271 on Build GRCh38
rs771002013 Chr.9: 35738356-35738356 on Build GRCh38
rs1011987148 Chr.9: 35738328-35738328 on Build GRCh38
rs970543034 Chr.9: 35738431-35738431 on Build GRCh38
rs748072207 Chr.9: 35738426-35738426 on Build GRCh38
rs749892510 Chr.9: 35738396-35738396 on Build GRCh38
rs746728147 Chr.9: 35738308-35738308 on Build GRCh38
rs769850454 Chr.9: 35738331-35738331 on Build GRCh38
rs765904160 Chr.9: 35738402-35738402 on Build GRCh38
rs751265524 Chr.9: 35738414-35738414 on Build GRCh38
rs754829622 Chr.9: 35738320-35738320 on Build GRCh38
rs757945302 Chr.9: 35738399-35738399 on Build GRCh38
rs773344585 Chr.9: 35738333-35738333 on Build GRCh38
rs754706242 Chr.9: 35738419-35738419 on Build GRCh38
rs989227836 Chr.9: 35738330-35738330 on Build GRCh38
rs759783962 Chr.9: 35738364-35738364 on Build GRCh38
rs749376850 Chr.9: 35738341-35738341 on Build GRCh38
rs781075917 Chr.9: 35738321-35738321 on Build GRCh38
rs761325530 Chr.9: 35738391-35738391 on Build GRCh38
rs764699917 Chr.9: 35738395-35738395 on Build GRCh38
rs781022451 Chr.9: 35738420-35738420 on Build GRCh38
rs750088946 Chr.9: 35738268-35738268 on Build GRCh38
rs748131725 Chr.9: 35738329-35738329 on Build GRCh38
rs751675685 Chr.9: 35738337-35738349 on Build GRCh38
rs947498456 Chr.9: 35738430-35738430 on Build GRCh38
rs776121281 Chr.9: 35738375-35738375 on Build GRCh38
rs764892022 Chr.9: 35738267-35738267 on Build GRCh38
rs202088139 Chr.9: 35738374-35738374 on Build GRCh38

View Full Product Details