Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 24719002-24719263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFNA5
Entrez Gene ID
1687
Gene Name
DFNA5, deafness associated tumor suppressor
Gene Aliases
ICERE-1
UniGene
Hs.520708
Gene Chromosome Location
Chr.7: 24698355-24758020 on Build GRCh38
Transcripts
NM_001127453.1 NM_001127454.1 NM_004403.2 XM_017011802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3637613 c.558C>T p.I186I Chr.7: 24719065-24719065 on Build GRCh38
1450108 c.473delT p.L158fs*1 Chr.7: 24719150-24719150 on Build GRCh38
6233313 c.478C>A p.Q160K Chr.7: 24719145-24719145 on Build GRCh38
6642775 c.528C>G p.V176V Chr.7: 24719095-24719095 on Build GRCh38
3762598 c.489G>A p.T163T Chr.7: 24719134-24719134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769548233 Chr.7: 24719089-24719089 on Build GRCh38
rs774826763 Chr.7: 24719025-24719025 on Build GRCh38
rs753880387 Chr.7: 24719115-24719115 on Build GRCh38
rs781327973 Chr.7: 24719137-24719137 on Build GRCh38
rs375535367 Chr.7: 24719197-24719197 on Build GRCh38
rs369866737 Chr.7: 24719202-24719202 on Build GRCh38
rs768321674 Chr.7: 24719239-24719239 on Build GRCh38
rs764265448 Chr.7: 24719200-24719200 on Build GRCh38
rs777636529 Chr.7: 24719132-24719132 on Build GRCh38
rs546120306 Chr.7: 24719143-24719143 on Build GRCh38
rs754008145 Chr.7: 24719042-24719042 on Build GRCh38
rs764515710 Chr.7: 24719023-24719023 on Build GRCh38
rs876307 Chr.7: 24719026-24719026 on Build GRCh38
rs376265514 Chr.7: 24719070-24719070 on Build GRCh38
rs375143165 Chr.7: 24719158-24719158 on Build GRCh38
rs876306 Chr.7: 24719102-24719102 on Build GRCh38
rs775344506 Chr.7: 24719091-24719091 on Build GRCh38
rs753992851 Chr.7: 24719107-24719108 on Build GRCh38
rs138741730 Chr.7: 24719095-24719095 on Build GRCh38
rs876657449 Chr.7: 24719053-24719053 on Build GRCh38
rs756084267 Chr.7: 24719035-24719035 on Build GRCh38
rs367730024 Chr.7: 24719034-24719034 on Build GRCh38
rs184980931 Chr.7: 24719054-24719054 on Build GRCh38
rs202246404 Chr.7: 24719230-24719230 on Build GRCh38
rs748849547 Chr.7: 24719234-24719234 on Build GRCh38
rs557149800 Chr.7: 24719127-24719127 on Build GRCh38
rs778475292 Chr.7: 24719240-24719240 on Build GRCh38
rs759658094 Chr.7: 24719120-24719120 on Build GRCh38
rs201217155 Chr.7: 24719022-24719022 on Build GRCh38
rs781275023 Chr.7: 24719222-24719222 on Build GRCh38
rs745848574 Chr.7: 24719076-24719076 on Build GRCh38
rs758262467 Chr.7: 24719134-24719135 on Build GRCh38
rs760983599 Chr.7: 24719164-24719164 on Build GRCh38
rs201176139 Chr.7: 24719038-24719038 on Build GRCh38
rs139931428 Chr.7: 24719154-24719154 on Build GRCh38
rs150265952 Chr.7: 24719047-24719047 on Build GRCh38
rs534323640 Chr.7: 24719051-24719051 on Build GRCh38
rs148449230 Chr.7: 24719167-24719167 on Build GRCh38
rs754555 Chr.7: 24719134-24719134 on Build GRCh38
rs367647197 Chr.7: 24719074-24719074 on Build GRCh38
rs143907715 Chr.7: 24719155-24719155 on Build GRCh38
rs979892116 Chr.7: 24719169-24719169 on Build GRCh38
rs763765525 Chr.7: 24719194-24719195 on Build GRCh38
rs147416797 Chr.7: 24719161-24719161 on Build GRCh38
rs766327140 Chr.7: 24719106-24719106 on Build GRCh38
rs541450984 Chr.7: 24719196-24719196 on Build GRCh38
rs770300688 Chr.7: 24719073-24719073 on Build GRCh38
rs544382620 Chr.7: 24719064-24719064 on Build GRCh38
rs768179307 Chr.7: 24719144-24719144 on Build GRCh38
rs371705826 Chr.7: 24719131-24719131 on Build GRCh38
rs761265129 Chr.7: 24719103-24719103 on Build GRCh38
rs762267830 Chr.7: 24719094-24719094 on Build GRCh38
rs876305 Chr.7: 24719176-24719176 on Build GRCh38
rs373591364 Chr.7: 24719031-24719031 on Build GRCh38
rs745411723 Chr.7: 24719079-24719079 on Build GRCh38
rs754554 Chr.7: 24719199-24719199 on Build GRCh38
rs754553 Chr.7: 24719233-24719233 on Build GRCh38
rs757058513 Chr.7: 24719204-24719204 on Build GRCh38
rs755221661 Chr.7: 24719043-24719043 on Build GRCh38

View Full Product Details