Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 18434795-18435052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDHC
Entrez Gene ID
3948
Gene Name
lactate dehydrogenase C
Gene Aliases
CT32, LDH3, LDHX
UniGene
Hs.654377
Gene Chromosome Location
Chr.11: 18412298-18451246 on Build GRCh38
Transcripts
NM_002301.4 NM_017448.3 XM_017017721.1 XM_017017722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1353045 c.503C>G p.A168G Chr.11: 18434824-18434824 on Build GRCh38
4404903 c.530A>G p.K177R Chr.11: 18434851-18434851 on Build GRCh38
428784 c.574G>A p.E192K Chr.11: 18434895-18434895 on Build GRCh38
6551821 c.534G>T p.L178F Chr.11: 18434855-18434855 on Build GRCh38
7296440 c.526G>T p.E176* Chr.11: 18434847-18434847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756713645 Chr.11: 18434885-18434885 on Build GRCh38
rs919663682 Chr.11: 18434949-18434949 on Build GRCh38
rs780275729 Chr.11: 18434919-18434919 on Build GRCh38
rs767205516 Chr.11: 18434850-18434850 on Build GRCh38
rs73438668 Chr.11: 18434989-18434989 on Build GRCh38
rs748279787 Chr.11: 18434947-18434947 on Build GRCh38
rs917602427 Chr.11: 18435000-18435000 on Build GRCh38
rs148817816 Chr.11: 18434826-18434826 on Build GRCh38
rs752749723 Chr.11: 18434854-18434854 on Build GRCh38
rs765184584 Chr.11: 18434938-18434938 on Build GRCh38
rs201168555 Chr.11: 18434832-18434832 on Build GRCh38
rs769056269 Chr.11: 18434939-18434939 on Build GRCh38
rs939843539 Chr.11: 18434982-18434983 on Build GRCh38
rs764039083 Chr.11: 18434868-18434868 on Build GRCh38
rs753372115 Chr.11: 18434878-18434878 on Build GRCh38
rs759454589 Chr.11: 18434842-18434842 on Build GRCh38
rs569599268 Chr.11: 18435008-18435008 on Build GRCh38
rs189886703 Chr.11: 18434909-18434909 on Build GRCh38
rs1007827759 Chr.11: 18434874-18434874 on Build GRCh38
rs909825269 Chr.11: 18434967-18434967 on Build GRCh38
rs1049951632 Chr.11: 18435014-18435014 on Build GRCh38
rs866403111 Chr.11: 18434828-18434828 on Build GRCh38
rs773537130 Chr.11: 18434833-18434833 on Build GRCh38
rs533091705 Chr.11: 18434970-18434970 on Build GRCh38
rs73438666 Chr.11: 18434964-18434964 on Build GRCh38
rs745495410 Chr.11: 18434900-18434900 on Build GRCh38
rs750492944 Chr.11: 18434869-18434869 on Build GRCh38
rs778572910 Chr.11: 18434898-18434898 on Build GRCh38
rs1044941244 Chr.11: 18435019-18435019 on Build GRCh38
rs973609247 Chr.11: 18434944-18434944 on Build GRCh38
rs756011979 Chr.11: 18434855-18434855 on Build GRCh38

View Full Product Details