Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 74843158-74843393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT6B
Entrez Gene ID
23522
Gene Name
lysine acetyltransferase 6B
Gene Aliases
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf
UniGene
Hs.35758
Gene Chromosome Location
Chr.10: 74824927-75032623 on Build GRCh38
Transcripts
NM_001256468.1 NM_001256469.1 NM_012330.3 XM_005269664.2 XM_017016000.1 XM_017016001.1 XM_017016002.1 XM_017016003.1 XM_017016004.1 XM_017016005.1 XM_017016006.1 XM_017016007.1 XM_017016008.1 XM_017016009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
685315 c.436C>A p.P146T Chr.10: 74843293-74843293 on Build GRCh38
1702652 c.307C>T p.R103C Chr.10: 74843164-74843164 on Build GRCh38
1297443 c.406C>G p.Q136E Chr.10: 74843263-74843263 on Build GRCh38
7136011 c.363G>A p.P121P Chr.10: 74843220-74843220 on Build GRCh38
3440601 c.470A>C p.K157T Chr.10: 74843327-74843327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750020725 Chr.10: 74843367-74843367 on Build GRCh38
rs760705600 Chr.10: 74843185-74843185 on Build GRCh38
rs776337217 Chr.10: 74843304-74843304 on Build GRCh38
rs775368710 Chr.10: 74843212-74843214 on Build GRCh38
rs562996276 Chr.10: 74843317-74843317 on Build GRCh38
rs778694892 Chr.10: 74843242-74843242 on Build GRCh38
rs377346332 Chr.10: 74843212-74843212 on Build GRCh38
rs746869008 Chr.10: 74843298-74843298 on Build GRCh38
rs35018386 Chr.10: 74843364-74843365 on Build GRCh38
rs201828896 Chr.10: 74843219-74843219 on Build GRCh38
rs370236468 Chr.10: 74843285-74843285 on Build GRCh38
rs993417725 Chr.10: 74843229-74843229 on Build GRCh38
rs779373950 Chr.10: 74843366-74843366 on Build GRCh38
rs771449717 Chr.10: 74843247-74843247 on Build GRCh38
rs1025323349 Chr.10: 74843334-74843334 on Build GRCh38
rs756912762 Chr.10: 74843241-74843241 on Build GRCh38
rs41314998 Chr.10: 74843232-74843232 on Build GRCh38
rs756668320 Chr.10: 74843220-74843220 on Build GRCh38
rs184609018 Chr.10: 74843331-74843331 on Build GRCh38
rs766588800 Chr.10: 74843204-74843204 on Build GRCh38
rs199812020 Chr.10: 74843218-74843218 on Build GRCh38
rs761606204 Chr.10: 74843309-74843309 on Build GRCh38
rs1016693351 Chr.10: 74843207-74843207 on Build GRCh38
rs778245309 Chr.10: 74843221-74843221 on Build GRCh38
rs763852432 Chr.10: 74843188-74843188 on Build GRCh38
rs776430152 Chr.10: 74843193-74843193 on Build GRCh38
rs769646911 Chr.10: 74843321-74843321 on Build GRCh38
rs143630427 Chr.10: 74843322-74843322 on Build GRCh38
rs147013911 Chr.10: 74843211-74843211 on Build GRCh38
rs764712157 Chr.10: 74843339-74843339 on Build GRCh38
rs763117455 Chr.10: 74843200-74843200 on Build GRCh38
rs1054836015 Chr.10: 74843282-74843282 on Build GRCh38
rs576505982 Chr.10: 74843361-74843361 on Build GRCh38
rs754453284 Chr.10: 74843342-74843342 on Build GRCh38
rs779662050 Chr.10: 74843280-74843280 on Build GRCh38
rs533770793 Chr.10: 74843243-74843243 on Build GRCh38
rs752855653 Chr.10: 74843332-74843332 on Build GRCh38

View Full Product Details